Skip to Content
Merck
All Photos(1)

Key Documents

I0775000

Isosorbide dinitrate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

ISDN

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8N2O8
CAS Number:
Molecular Weight:
236.14
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

isosorbide

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[H][C@@]1([C@@]([H])(O[N+]([O-])=O)CO2)[C@@]2([H])[C@]([H])(O[N+]([O-])=O)CO1

InChI

1S/C6H8N2O8/c9-7(10)15-3-1-13-6-4(16-8(11)12)2-14-5(3)6/h3-6H,1-2H2

InChI key

MOYKHGMNXAOIAT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Isosorbide dinitrate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Desen. Expl. 3

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Giuseppe Di Gioia et al.
European heart journal, 41(17), 1665-1672 (2019-08-17)
Fractional flow reserve (FFR) has never been investigated in patients with reduced ejection fraction and associated coronary artery disease (CAD). We evaluated the impact of FFR on the management strategies of these patients and related outcomes. From 2002 to 2010
Eriko Maeda et al.
SpringerPlus, 4, 697-697 (2015-11-21)
To evaluate the degree of heart rate (HR) changes at rest (HRrest), during breath hold (HRtest), and during cardiac CT examinations (HRscan) in a large group of patients , and to derive and asses the feasibility of a predictive formula
Stephane Fournier et al.
Journal of the American Heart Association, 9(24), e017729-e017729 (2020-12-08)
Background Global fractional flow reserve (FFR) (ie, the sum of the FFR values in the 3 major coronary arteries) is a physiologic correlate of global atherosclerotic burden. The objective of the present study was to investigate the value of global
Ryo Kurokawa et al.
Medicine, 98(19), e15538-e15538 (2019-05-15)
To compare coronary artery luminal enhancement in coronary computed tomography angiography (CCTA) between ventral and dorsal region-of-interest (ROI) bolus tracking in the descending aorta.The records of 165 consecutive patients who underwent CCTA with non-helical acquisition from July 2017 to March
S Silber
European journal of clinical pharmacology, 38 Suppl 1, S35-S51 (1990-01-01)
Nitrates are highly effective both in terminating acute attacks of angina pectoris and in the prophylaxis of symptomatic and asymptomatic myocardial ischemia. Preload reduction by venodilatation is the prevailing mechanism of nitrates in patients with chronic stable angina and is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service