Skip to Content
Merck
All Photos(1)

Key Documents

1466506

USP

Nitroglycerin in propylene glycol

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Glycerol trinitrate solution, Diluted Nitroglycerin, Diluted glyceryl trinitrate, 1,2,3-Propanetriol 1,2,3-trinitrate solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H5N3O9
Molecular Weight:
227.09
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

nitroglycerin

form

solution

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

single component solution

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Nitroglycerin in propylene glycol USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Diluted Nitroglycerin
  • Nitroglycerin Injection

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

related product

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

217.4 °F - closed cup

Flash Point(C)

103 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service