Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
grade
pharmaceutical primary standard
agency
USP
API family
isosorbide
manufacturer/tradename
USP
application(s)
pharmaceutical (small molecule)
format
neat
General description
Application
Also, for use with USP monographs such as:
- Isosorbide Mononitrate Extended-Release Tablets
- Isosorbide Mononitrate Tablets
- Isosorbide Dinitrate Extended-Release Tablets
Analysis Note
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| description United States Pharmacopeia (USP) Reference Standard | description European Pharmacopoeia (EP) Reference Standard | description United States Pharmacopeia (USP) Reference Standard | description United States Pharmacopeia (USP) Reference Standard |
| manufacturer/tradename USP | manufacturer/tradename EDQM | manufacturer/tradename USP | manufacturer/tradename USP |
| grade pharmaceutical primary standard | grade pharmaceutical primary standard | grade pharmaceutical primary standard | grade pharmaceutical primary standard |
| application(s) pharmaceutical (small molecule) | application(s) pharmaceutical (small molecule) | application(s) pharmaceutical (small molecule) | application(s) pharmaceutical (small molecule) |
| format neat | format neat | format neat | format neat |
| agency USP | agency - | agency USP | agency USP |
| API family isosorbide | API family isosorbide | API family isomalt | API family digitoxin |
signalword
Danger
hcodes
Hazard Classifications
Flam. Sol. 1
Storage Class
4.1B - Flammable solid hazardous materials
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


