Skip to Content
Merck
All Photos(1)

Key Documents

Y0001635

Diclofenac for system suitability

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Diclofenac sodium salt, 2-[(2,6-Dichlorophenyl)amino]benzeneacetic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H10Cl2NNaO2
CAS Number:
Molecular Weight:
318.13
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

diclofenac

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[Na+].[O-]C(=O)Cc1ccccc1Nc2c(Cl)cccc2Cl

InChI

1S/C14H11Cl2NO2.Na/c15-10-5-3-6-11(16)14(10)17-12-7-2-1-4-9(12)8-13(18)19;/h1-7,17H,8H2,(H,18,19);/q;+1/p-1

InChI key

KPHWPUGNDIVLNH-UHFFFAOYSA-M

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Diclofenac for system suitability EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Standard NSAID and cyclooxygenase (COX) inhibitor. Major metabolites are 4´-hydroxydiclofenac and 5´-hydroxydiclofenac. Has been used as substrate selective for CYP2C9.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 2 - Repr. 2 - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paul L McCormack et al.
Drugs, 68(1), 123-130 (2007-12-18)
*A new formulation of the nonselective NSAID diclofenac sodium suitable for intravenous bolus injection has been developed using hydroxypropyl beta-cyclodextrin as a solubility enhancer (HPbetaCD diclofenac). * HPbetaCD diclofenac intravenous bolus injection was shown to be bioequivalent to the existing
P A Todd et al.
Drugs, 35(3), 244-285 (1988-03-01)
Diclofenac is a non-steroidal anti-inflammatory drug (NSAID) advocated for use in painful and inflammatory rheumatic and certain non-rheumatic conditions. It is available in a number of administration forms which can be given orally, rectally or intramuscularly. Conveniently, dosage adjustments are
George M Martin et al.
Journal of drugs in dermatology : JDD, 11(5), 600-608 (2012-04-25)
Diclofenac sodium 3% gel (Solaraze®) gained US approval for the treatment of actinic keratosis (AK) more than 10 years ago. Since the publication of the pivotal phase 3 studies, numerous clinical studies have assessed use of this therapy in a
D Aygün et al.
Histology and histopathology, 27(4), 417-436 (2012-03-01)
Nonsteroidal anti-inflammatory drugs (NSAIDs) are widely used for the purpose of anti-inflammation, antipyretic, and analgesia. For this aim, they are used for the alleviation of pain, fever, and inflammation associated with rheumatoid arthritis, sports injuries, and temporary pain. However, treatment
Ryoichi Sawazaki et al.
Investigative ophthalmology & visual science, 55(4), 2547-2556 (2014-03-29)
Dry eye syndrome (DES) is characterized by an increase in tear osmolarity and induction of the expression and nuclear localization of an osmoprotective transcription factor (nuclear factor of activated T-cells 5 [NFAT5]) that plays an important role in providing protection

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service