Skip to Content
Merck

Skip To

21097

Calcium chloride dihydrate

BioUltra, Molecular Biology, ≥99.5% (KT)

Synonym(s):

CaCl2.2H2O

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
CaCl2 · 2H2O
CAS Number:
Molecular Weight:
147.01
NACRES:
NA.26
PubChem Substance ID:
UNSPSC Code:
12352302
EC Number:
233-140-8
MDL number:
Assay:
≥99.5% (KT)
Grade:
Molecular Biology
Form:
solid
Solubility:
H2O: 1 M at 20 °C, clear, colorless
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

Molecular Biology

Quality Segment

product line

BioUltra

assay

≥99.5% (KT)

form

solid

impurities

DNases, none detected, RNases, none detected, insoluble matter, passes filter test, phosphatases, none detected, proteases, none detected, ≤0.002% total nitrogen (N)

pH

5.0-8.0 (25 °C, 1 M in H2O)

mp

176 °C (dec.) (lit.)

solubility

H2O: 1 M at 20 °C, clear, colorless

anion traces

sulfate (SO42-): ≤50 mg/kg

cation traces

As: ≤0.1 mg/kg, Ba: ≤5 mg/kg, Bi: ≤5 mg/kg, Cd: ≤5 mg/kg, Co: ≤5 mg/kg, Cr: ≤5 mg/kg, Cu: ≤5 mg/kg, Fe: ≤5 mg/kg, K: ≤50 mg/kg, Li: ≤20 mg/kg, Mg: ≤50 mg/kg, Mn: ≤5 mg/kg, Mo: ≤5 mg/kg, Na: ≤100 mg/kg, Ni: ≤5 mg/kg, Pb: ≤5 mg/kg, Sr: ≤100 mg/kg, Zn: ≤5 mg/kg

λ

1 M in H2O

UV absorption

λ: 260 nm Amax: 0.015, λ: 280 nm Amax: 0.012

SMILES string

O.O.Cl[Ca]Cl

InChI

1S/Ca.2ClH.2H2O/h;2*1H;2*1H2/q+2;;;;/p-2

InChI key

LLSDKQJKOVVTOJ-UHFFFAOYSA-L

Application


  • Corncob-derived biodegradable packaging films: A sustainable solution for raspberry post-harvest preservation.: This study explores the use of corncob-derived biodegradable films infused with calcium chloride dihydrate to preserve raspberries post-harvest. The films effectively reduce microbial growth and extend the shelf life of the fruit (Paudel et al., 2024).

  • The Effect of a Setting Accelerator on the Physical and Mechanical Properties of a Fast-set White Portland Cement Mixed with Nano-zirconium Oxide.: The study examines the impact of calcium chloride dihydrate as a setting accelerator in cement composites, enhancing their mechanical properties and setting time, which is vital for construction applications (Chen et al., 2023).

  • Co-administration of Intravenous Drugs: Rapidly Troubleshooting the Solid Form Composition of a Precipitate in a Multi-drug Mixture Using On-Site Raman Spectroscopy.: This paper highlights the use of calcium chloride dihydrate in pharmaceutical formulations to stabilize drug mixtures and prevent precipitation, ensuring the efficacy of intravenous drugs (Nilsson et al., 2023).

  • Calcium chloride dihydrate supplementation at ICSI improves fertilization and pregnancy rates in patients with previous low fertilization: a retrospective paired treatment cycle study.: The research demonstrates that calcium chloride dihydrate supplementation during intracytoplasmic sperm injection (ICSI) significantly improves fertilization and pregnancy rates in patients with previous fertilization failures (Popkiss et al., 2022).

Other Notes

In the calcium phosphate method for the transfection of eukaryotic cells

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
208291C7902C3881
form

solid

form

solid

form

powder

form

powder

solubility

H2O: 1 M at 20 °C, clear, colorless

solubility

water: 150 mg/mL

solubility

H2O: 294 mg/mL, clear, colorless

solubility

-

assay

≥99.5% (KT)

assay

≥99% (complexometric)

assay

≥99.0%

assay

≥99.0%

Quality Level

200

Quality Level

100

Quality Level

200

Quality Level

200

grade

Molecular Biology

grade

Molecular Biology

grade

-

grade

-

impurities

DNases, none detected, insoluble matter, passes filter test, proteases, none detected, RNases, none detected, ≤0.002% total nitrogen (N), phosphatases, none detected

impurities

-

impurities

-

impurities

-


Still not finding the right product?

Explore all of our products under Calcium chloride dihydrate


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

13 - Non Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters