Skip to Content
Merck
All Photos(1)

Key Documents

C0691000

Ceftriaxone sodium

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Ceftriaxone disodium salt hemi(heptahydrate)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H16N8Na2O7S3 · 3.5H2O
CAS Number:
Molecular Weight:
661.60
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

ceftriaxone

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

O.O.O.O.O.O.O.[Na+].[Na+].CO\N=C(/C(=O)N[C@H]1C2SCC(CSC3=NC(=O)C(=O)N([Na])N3C)=C(N2C1=O)C([O-])=O)c4csc(N)n4.CO\N=C(/C(=O)N[C@H]5C6SCC(CSC7=NC(=O)C(=O)N([Na])N7C)=C(N6C5=O)C([O-])=O)c8csc(N)n8

InChI

1S/2C18H18N8O7S3.4Na.7H2O/c2*1-25-18(22-12(28)13(29)23-25)36-4-6-3-34-15-9(14(30)26(15)10(6)16(31)32)21-11(27)8(24-33-2)7-5-35-17(19)20-7;;;;;;;;;;;/h2*5,9,15H,3-4H2,1-2H3,(H5,19,20,21,23,27,29,31,32);;;;;7*1H2/q;;4*+1;;;;;;;/p-4/b2*24-8-;;;;;;;;;;;/t2*9-,15-;;;;;;;;;;;/m11.........../s1

InChI key

PMRZKYOXTPBIQF-MAODNAKNSA-J

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Ceftriaxone sodium EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Filippo Lagi et al.
Journal of travel medicine, 20(2), 128-130 (2013-03-08)
We present the case of two Australian tourists aged 25 and 26  years who, after immersion in a canal in Venice, developed severe leptospirosis. After a 1-week history of fever, headache, myalgia, and vomiting they developed jaundice and renal failure.
Catherine A Ison et al.
The Lancet. Infectious diseases, 13(9), 762-768 (2013-06-15)
Effective treatment of gonorrhoea is fundamental to public health control; however, the ability of Neisseria gonorrhoeae to successively develop resistance to different treatments has hampered control efforts. The extended-spectrum cephalosporins--cefixime and ceftriaxone--have been recommended in the UK for treatment of
Zhihui Luo et al.
Journal of biomedical nanotechnology, 9(1), 69-76 (2013-05-01)
In this paper, the selectively enhanced antibacterial effects of ZnO nanorods with several kinds of conventional medical antibiotics are investigated. Compares to gentamicin, clarithromycin and ofloxacin, ZnO nanorods could obviously achieve synergistic antibacterial effects with ceftriaxone against Escherichia coli (E.
Johan Chanal et al.
Presse medicale (Paris, France : 1983), 42(4 Pt 1), 454-458 (2013-02-26)
Bacterial culture remains the gold standard for symptomatic infection. Nucleic Acid Amplification Tests (NAATs) have better sensitivity and specificity for rectal and pharyngeal specimens. A bacterial culture with antibiogram must be done for all NAAT positive specimens in order to
Hemalatha Srinivasalu et al.
Pediatrics, 131(6), e1977-e1981 (2013-05-15)
Erythema of the ear lobe in the context of Lyme disease is caused by either borrelial lymphocytoma or localized erythema migrans. Here we present a case of chondritis limited to the ear cartilage caused by Lyme disease. The patient was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service