Skip to Content
Merck
All Photos(1)

Key Documents

PHR1382

Supelco

Ceftriaxone Sodium

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Ceftriaxone disodium salt hemi(heptahydrate)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H16N8Na2O7S3 · 3.5H2O
CAS Number:
Molecular Weight:
661.60
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to BP 976
traceable to Ph. Eur. C0691000
traceable to USP 1098184

API family

ceftriaxone

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O.O.O.O.O.O.O.[Na+].[Na+].CO\N=C(/C(=O)N[C@H]1C2SCC(CSC3=NC(=O)C(=O)N([Na])N3C)=C(N2C1=O)C([O-])=O)c4csc(N)n4.CO\N=C(/C(=O)N[C@H]5C6SCC(CSC7=NC(=O)C(=O)N([Na])N7C)=C(N6C5=O)C([O-])=O)c8csc(N)n8

InChI

1S/2C18H18N8O7S3.4Na.7H2O/c2*1-25-18(22-12(28)13(29)23-25)36-4-6-3-34-15-9(14(30)26(15)10(6)16(31)32)21-11(27)8(24-33-2)7-5-35-17(19)20-7;;;;;;;;;;;/h2*5,9,15H,3-4H2,1-2H3,(H5,19,20,21,23,27,29,31,32);;;;;7*1H2/q;;4*+1;;;;;;;/p-4/b2*24-8-;;;;;;;;;;;/t2*9-,15-;;;;;;;;;;;/m11.........../s1

InChI key

PMRZKYOXTPBIQF-MAODNAKNSA-J

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

Ceftriaxone Sodium may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations and serum samples by various chromatography techniques.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC3447 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Validation of an HPLC-UV method for the determination of ceftriaxone sodium residues on stainless steel surface of pharmaceutical manufacturing equipments
Akl MA, et al.
Journal of Pharmaceutical and Biomedical Analysis, 55(2), 247-252 (2011)
Simultaneous Determination of Ceftriaxone Sodium and Non Steroidal Anti?Inflammatory Drugs in Pharmaceutical Formulations and Human Serum by RP?HPLC.
Sultana N, et al.
J. Chin. Chem. Soc., 57(6), 1278-1285 (2010)
Simultaneous determination of ceftriaxone sodium and statin drugs in pharmaceutical formulations and human serum by RP-HPLC.
Sultana N, et al.
Journal of the Chilean Chemical Society, 55(2), 193-198 (2010)
Michael S Northrop et al.
Journal of pediatric hematology/oncology, 37(1), e63-e66 (2014-06-01)
Ceftriaxone is a frequently used empiric antibiotic in children. Acute hemolysis is a rare side effect of ceftriaxone therapy associated with a high mortality rate. A 14-year-old boy suffering from Crohn disease developed bacterial pneumonia that was treated with ceftriaxone.
Scott V Monte et al.
Expert opinion on drug safety, 7(5), 515-523 (2008-09-02)
Isolated reports of neonatal and infant deaths associated with ceftriaxone-calcium precipitation in the lungs and kidneys have prompted a recommendation from the US FDA in June 2007 advising that in patients of all ages, calcium-containing solutions should not be administered

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service