Skip to Content
Merck
All Photos(1)

Key Documents

B0490000

Benzarone

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

(2-Ethylbenzofuran-3-yl)(4-hydroxyphenyl)methanone, NSC 82134

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H14O3
CAS Number:
Molecular Weight:
266.29
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

benzarone

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[o]1c2c(c(c1CC)C(=O)c3ccc(cc3)O)cccc2

InChI

1S/C17H14O3/c1-2-14-16(13-5-3-4-6-15(13)20-14)17(19)11-7-9-12(18)10-8-11/h3-10,18H,2H2,1H3

InChI key

RFRXIWQYSOIBDI-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Benzarone EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W Voelter et al.
Journal of chromatography, 290, 1-12 (1984-05-04)
Using 2-ethyl-3-(4- hydroxybenzoyl )benzofuran ( EHBB ) as an example, biotransformation in rabbits and rats and by rat hepatocyte suspensions was studied by high-performance liquid chromatography (HPLC) and mass spectrometry. The biotransformation of N-alkyl-substituted piperidines by rat hepatocytes gives valuable
H Iven et al.
Cancer chemotherapy and pharmacology, 21(4), 337-342 (1988-01-01)
In anesthetized rabbits, continuous infusion of methotrexate (MTX; 30 micrograms kg-1 min-1) established steady-state plasma concentrations for MTX and the metabolite 7-hydroxymethotrexate (7-OH-MTX) within 40 min. Fifty percent of the infused dose was eliminated unchanged by the kidneys and the
[On the site of action of benzarone and some other agents influencing the intracellular concentration of cyclic nucleotides].
L O Zwillenberg et al.
Arzneimittel-Forschung, 32(9), 1114-1120 (1982-01-01)
Subfulminant hepatitis requiring liver transplantation after benzarone administration.
M Gehenot et al.
Journal of hepatology, 20(6), 842-842 (1994-06-01)
J X de Vries et al.
Journal of chromatography, 382, 167-174 (1986-10-31)
Benzarone (the debrominated metabolite of the uricosuric drug benzbromarone) has been proposed for treatment of vascular disorders. An assay was developed for the quantitation of total benzarone (conjugated and unconjugated) in plasma and urine, following oral intake of benzarone. Enzymatic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service