Skip to Content
Merck
All Photos(1)

Documents

C-036

Supelco

Chlorpheniramine maleate salt solution

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

Chlorpheniramine maleate solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H19ClN2 · C4H4O4
CAS Number:
Molecular Weight:
390.86
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

SNAP-N-SPIKE®, SNAP-N-SHOOT®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

pharmaceutical (small molecule)

format

single component solution

storage temp.

−20°C

InChI

1S/C16H19ClN2.C4H4O4/c1-19(2)12-10-15(16-5-3-4-11-18-16)13-6-8-14(17)9-7-13;5-3(6)1-2-4(7)8/h3-9,11,15H,10,12H2,1-2H3;1-2H,(H,5,6)(H,7,8)/b;2-1-

InChI key

DBAKFASWICGISY-BTJKTKAUSA-N

Gene Information

human ... HRH1(3269)

General description

An analytical reference standard suitable for use as calibrators or controls in a variety of LC/MS or GC/MS applications such as clinical toxicology or pharmaceutical research. Chlorphenamine is a first-generation antihistamine prescribed for the prevention of allergic conditions such as rhinitis.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anaphylactic reaction to ranitidine and dexchlorpheniramine.
C Thurot-Guillou et al.
European journal of dermatology : EJD, 17(2), 170-171 (2007-03-06)
Eef L Theunissen et al.
British journal of clinical pharmacology, 61(1), 79-86 (2006-01-05)
Previous studies have demonstrated that the antihistamines mequitazine, cetirizine and dexchlorpheniramine produce mild sedation after single doses. It is unknown, however, whether acute sedation persists after repeated dosing. Therefore, this study assessed the effects of repeated dosing of these antihistamines
Urticaria due to aloe vera: a new sensitizer?
Alberto Alvarez-Perea et al.
Annals of allergy, asthma & immunology : official publication of the American College of Allergy, Asthma, & Immunology, 105(5), 404-405 (2010-11-09)
O Cáceres Calle et al.
Allergologia et immunopathologia, 32(5), 306-309 (2004-10-01)
Dexchlorpheniramine (DH) is a classical or first generation antihistamine belonging to the ethanolamine group. Adverse effects related to these antihistamines are frequent, but the hypersensitivity reactions described in the literature since 1940 are exceptional. We report the case of a
P van Ruitenbeek et al.
Journal of psychopharmacology (Oxford, England), 22(6), 663-672 (2008-01-23)
Results from recent animal studies suggest an important role for histamine in memory functioning. Histaminergic drugs might prove beneficial for people suffering from memory impairment. To determine if histamine is involved in memory functioning this study evaluates the effects of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service