Skip to Content
Merck

IX0185

Iodomethane

Synonym(s):

Iodomethane, Methyl Iodide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
CH3I
CAS Number:
Molecular Weight:
141.94
UNSPSC Code:
12352101
EC Index Number:
200-819-5
MDL number:
NACRES:
NA.21
Assay:
≥98% (GC)
Bp:
42 °C/1013 hPa
Vapor pressure:
540 hPa ( 25 °C)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

540 hPa ( 25 °C)

Quality Segment

assay

≥98% (GC)

form

liquid

color

APHA: ≤100

pH

5.2 (25 °C in H2O)

bp

42 °C/1013 hPa

mp

-66 °C

density

2.28 g/cm3 at 20 °C

shipped in

ambient

storage temp.

room temp

SMILES string

IC

InChI

1S/CH3I/c1-2/h1H3

InChI key

INQOMBQAUSQDDS-UHFFFAOYSA-N

General description

EMPARTA solvents are designed for routine chemical synthesis, drying, purification, and critical labware cleaning in quality control environments. Engineered for precision, they deliver consistently reproducible results essential for rigorous validation and documentation.

Exceptional Quality Standards: These solvents meet or exceed ACS specifications, providing assurance of high quality that is crucial for accurate testing and analysis in quality control.

Consistent Reproducibility: Designed for precision, the solvents ensure that results can be reliably reproduced, which is vital for maintaining compliance and integrity in quality control processes.

Versatile Applications: Suitable for a variety of QC tasks, including analytical testing, method validation, and testing USP/NF monographs, these solvents support a comprehensive approach to quality assurance in laboratory settings.

Application

IX0185-3 is packaged in poly coated glass bottles

Analysis Note

Assay (GC): 98% min
Color (APHA): 100APHA max
Form: Clear liquid
Infrared Spectrum: Conforms to standard


Still not finding the right product?

Explore all of our products under Iodomethane


Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 2 - Carc. 2 - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class

3 - Flammable liquids

WGK

WGK 3

Flash Point (°F)

89.8 °F - closed cup

Flash Point (°C)

32.1 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library