Skip to Content
Merck

8.51209

D(+)-Biotin

Novabiochem®

Synonym(s):

D(+)-Biotin, D-Biotin,Vitamin H, Hexahydro-2-oxothieno(3,4-d)-imidazole-4-pentanoic acid, Vitamin H

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C10H16N2O3S
CAS Number:
Molecular Weight:
244.31
UNSPSC Code:
41116133
EC Index Number:
200-399-3
NACRES:
NA.22
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

product line

Novabiochem®

form

solid

potency

>2000 mg/kg LD50, oral (Rat)

reaction suitability

reaction type: Biotinylations

manufacturer/tradename

Novabiochem®

pH

4.5 (0.1 g/L in H2O)

mp

232-233 °C

solubility

0.2 g/L

bulk density

200‑500 kg/m3

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

S1C(C2NC(=O)NC2C1)CCCCC(=O)O

InChI

1S/C10H16N2O3S/c13-8(14)4-2-1-3-7-9-6(5-16-7)11-10(15)12-9/h6-7,9H,1-5H2,(H,13,14)(H2,11,12,15)

InChI key

YBJHBAHKTGYVGT-UHFFFAOYSA-N

General description

D(+)-Biotin is the simplest reagent for the introduction of biotin to peptides, proteins and other biomolecules. In SPPS, it is best coupled using DIPCDI/Oxyma Pure. Biotin-labeled peptides have many important applications in immunology and histochemistry, such as affinity purification and FRET-based flow cytometry, solid-phase immunoassays, and receptor localization, that exploit the high affinity of streptavidin and avidin for biotin.,,,

Associated Protocols and Technical Articles
Biotinylation Reagents for Peptide Synthesis

Literature references:
[1] K. Hofmann & Y. Kiso (1976) Proc. Natl. Acad. Sci., 73, 3516.
[2] T. Buranda, et al. (1999) Cytometry, 37, 21.
[3] I. Sélo, et al. (1996) J. Immunol. Methods, 199, 127.
[4] J. Howl, et al. (1993) Eur J Biochem., 213, 711.

Application

Some recent applications of D(+)-Biotin in SPPS:
  • In the high-throughput biotinylation of proteins. The approach for site-specific biotinylation of proteins is useful for various applications such as protein purification and analysis.
  • In the site-specific biotinylation of multipodal glycoclusters on a solid support. The synthesis of biotinylated glycoclusters, has important applications in glycobiology.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under D(+)-Biotin


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Fluorescent and biotinylated linear peptides as selective bifunctional ligands for the V1a vasopressin receptor
J. Howl, et al.
European Journal of Biochemistry, 213, 711-711 (1993)
An approach to the targeted attachment of peptides and proteins to solid supports.
K. Hofmann & Y. Kiso
Proceedings of the National Academy of Sciences, 73, 3516-3516 (1976)
Synthesis of biotinylated multipodal glycoclusters on a solid support
Karskela M, et al.
European Journal of Organic Chemistry, 2012, 6594-6605 (2012)



Global Trade Item Number

SKUGTIN
851209000504027269341404
851209999904027269341343
851209000104027269341374
851209000004027269341312
851209002504027269341435