Skip to Content
Merck
All Photos(1)

Key Documents

N4382

Sigma-Aldrich

Nalidixic acid sodium salt

powder

Synonym(s):

1-Ethyl-1,4-dihydro-7-methyl-4-oxo-1,8-naphthyridine-3-carboxylic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H11N2NaO3
CAS Number:
Molecular Weight:
254.22
Beilstein:
3580062
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.77

form

powder

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

Mode of action

DNA synthesis | interferes
enzyme | inhibits

originator

Sanofi Aventis

SMILES string

[Na+].CCN1C=C(C([O-])=O)C(=O)c2ccc(C)nc12

InChI

1S/C12H12N2O3.Na/c1-3-14-6-9(12(16)17)10(15)8-5-4-7(2)13-11(8)14;/h4-6H,3H2,1-2H3,(H,16,17);/q;+1/p-1

InChI key

ROKRAUFZFDQWLE-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: quinolone
Nalidixic acid is a quinolone and can block the activity of bacterial topoisomerase type II enzyme, thereby stopping bacterial replication. It is a potential antibacterial agent against Gram-negative bacteria than Gram-positive bacteria. Nalidixic acid has been studied in the management of urinary tract infections.

Application

Nalidixicacid has been used to treat the bacterial cultures to study itsresistance on Salmonella enterica and SalmonellaTyphimurium.

Biochem/physiol Actions

Nalidixic acid sodium salt is an inhibitor of bacterial DNA polymerase (DNA gyrase) and avian myeloblastoma virus reverse transcriptase. Inhibits nucleic acid and protein synthesis in Saccharomyces cerevisiae.

Features and Benefits

This compound is a featured product for Apoptosis research. Click here to discover more featured Apoptosis products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound was developed by Sanofi Aventis. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Daniel Hernandez-Patlan et al.
Frontiers in veterinary science, 6, 108-108 (2019-05-21)
Decreases in the use of antibiotics and anticoccidials in the poultry industry have risen the appearance of necrotic enteritis (NE). The purpose of this study was to evaluate the effect of a Bacillus direct-fed microbial (DFM) on growth performance, intestinal
Nalidixic Acid
Acta Entomologica Silesiana, 154-175 (1979)
Thu D M Pham et al.
MedChemComm, 10(10), 1719-1739 (2019-12-06)
The quinolone antibiotics arose in the early 1960s, with the first examples possessing a narrow-spectrum of activity with unfavorable pharmacokinetic properties. Over time, the development of new quinolone antibiotics has led to improved analogues with an expanded spectrum and high
Omar F Prado-Rebolledo et al.
Avian pathology : journal of the W.V.P.A, 46(1), 90-94 (2016-08-23)
Two experiments were conducted to evaluate the effect of a lactic acid bacteria-based probiotic (FloraMax-B11®) against Salmonella enterica serovar Enteritidis intestinal colonization and intestinal permeability in broiler chickens. Experiment 1 consisted of two independent trials. In each trial, day-old broiler
Margarita A Arreguin-Nava et al.
Animals : an open access journal from MDPI, 9(9) (2019-09-20)
The effect of Lactobacillus spp.-based probiotic candidates on Salmonella enterica serovar Enteritidis (SE) colonization was evaluated in two separate experiments. In each experiment, sixty-one day-of-hatch female turkey poults were obtained from a local hatchery. In both experiments, poults were challenged

Articles

Cell cycle phases (G1, S, G2, M) regulate cell growth, DNA replication, and division in proliferating cells.

Cell cycle phases (G1, S, G2, M) regulate cell growth, DNA replication, and division in proliferating cells.

Cell cycle phases (G1, S, G2, M) regulate cell growth, DNA replication, and division in proliferating cells.

Cell cycle phases (G1, S, G2, M) regulate cell growth, DNA replication, and division in proliferating cells.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service