Skip to Content
Merck
All Photos(1)

Key Documents

95577

Sigma-Aldrich

Chelex® 100 sodium form

100-200 mesh particle size

Synonym(s):

crosslinked polystyrene-iminodiacetate ion exchange resin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
23151817
NACRES:
NA.22

product line

Chelex®

Quality Level

form

crystals

reaction suitability

reaction type: solution phase peptide synthesis

loss

65-85% loss on drying, 110°C

matrix

polystyrene/DVB

matrix active group

paired iminodiacetate groups

particle size

100-200 mesh

Looking for similar products? Visit Product Comparison Guide

General description

Chelex®100 sodium form resin is a styrene divinylbenzene copolymer with a pair of iminodiacetate ions that serve as chelating groups to bind polyvalent metal ions.

Application

Chelex®100 can be used:
  • As an ion exchange resin in wastewater treatment it has a high affinity for metal ions and selectivity for transition metals such as Zn, Co, Ni, and Cu
  • In the extraction and purification of DNA

Legal Information

Chelex is a registered trademark of Bio-Rad Laboratories, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Real-time PCR for detection of Trypanosoma brucei in human blood samples
S Becker, et al.
Diagnostic Microbiology and Infectious Disease, 50, 193-199 (2004)
All-in-one microfluidic nucleic acid diagnosis system for multiplex detection of sexually transmitted pathogens directly from genitourinary secretions
X Ye, et al.
Talanta, 221, 121462-121462 (2021)
Separation of uranium from natural waters on chelex-100 resin.
Pakalns P.
Analytica Chimica Acta, 120, 289-296 (1980)
Layman's guide to common complaints. 11. Food poisoning.
I Librach
Nursing mirror, 149(13), 24-25 (1979-09-27)
Removal of trace metals from marine culture media.
Davey EW, et al.
Limnology and Oceanography, 15(3), 486-488 (1970)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service