Skip to Content
Merck
All Photos(4)

Key Documents

901874

Sigma-Aldrich

Ammonium sulfate

anhydrous, Redi-Dri, ReagentPlus®, ≥99.0%

Synonym(s):

Mascagnite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH4)2SO4
CAS Number:
Molecular Weight:
132.14
MDL number:
UNSPSC Code:
12352116
NACRES:
NA.21
Assay:
≥99.0%
grade:
anhydrous
form:
powder
solubility:
water: soluble 132 g/L at 20 °C
Pricing and availability is not currently available.

grade

anhydrous

Quality Level

product line

ReagentPlus®
Redi-Dri

Assay

≥99.0%

form

powder

pH

5-6 (25 °C, 132 g/L)

mp

>280 °C (dec.) (lit.)

solubility

water: soluble 132 g/L at 20 °C

density

1.77 g/mL at 25 °C (lit.)

cation traces

Fe: ≤10 ppm
Pb: ≤3 ppm
Se: ≤5 ppm

SMILES string

N.N.OS(O)(=O)=O

InChI

1S/2H3N.H2O4S/c;;1-5(2,3)4/h2*1H3;(H2,1,2,3,4)

InChI key

BFNBIHQBYMNNAN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Ammonium sulfate is an ammonium salt commonly used as fertilizer. It can be synthesized by neutralizing sulfuric acid with ammonia.[1] It is reported to enhance the phytotoxicity of several herbicides like glyphosates.[2] It is formed as a co-product during the manufacture of caprolactam, a precursor of nylon.[3]

Redi-Dri technology eliminates clumping and the need for complicated procedures to keep your salts free-flowing. Keeping your salts dry helps improve your results by giving accurate measurements and easier-to-handle salts and buffers. Rather than drying your salts in an oven or desiccator or breaking them apart manually, Redi-Dri ensures that your salts will remain easy-to-use for months without worrying about anti-caking agents. Even after repeated openings you can be assured that your results are always accurate. Redi-Dri saves you time, energy and money by keeping your salts usable for longer!

Application

Ammonium sulfate may be used in methyl cellulose precipitable tannin assay to examine wine tannin in finished wine samples.[4] It may also be used in making externally mixed aerosols for analyzing the 1 × 3 tandem differential mobility analyzer (1 × 3-TDMA), which is mainly employed for determining the irreversibility of the hygroscopic growth factor of aerosol particles.[5]

Features and Benefits

  • Increased efficiency – Eliminate time and effort of loosening hardened powders
  • Safety –Avoid risks of using tools of force to break up clumps
  • Economy –Faster preparation and solvation increase productivity and reduce costs
  • Assured quality –Excellent, expert-tested quality with no anti-caking agents
  • Flexible volumes –Available from research amounts to scale-up quantities

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany
Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The 1-by-3 tandem differential mobility analyzer for measurement of the irreversibility of the hygroscopic growth factor.
Rosenoern T, et al.
Aerosol Science and Technology, 43(7), 641-652 (2009)
Influence of yeast strain on Shiraz wine quality indicators.
Holt H, et al.
International Journal of Food Microbiology, 165(3), 302-311 (2013)
Industrial catalytic aspects of the synthesis of monomers for nylon production.
Bellussi G and Perego C.
CATTECH, 4(1), 4-16 (2000)
Ammonium Compounds.
Weston CW, et al.
Kirk-Othmer Encyclopedia of Chemical Technology (2003)
Simon Gareth Broadley et al.
Acta crystallographica. Section D, Biological crystallography, 68(Pt 11), 1450-1459 (2012-10-24)
MshB, a zinc-based deacetylase, catalyses a step in the mycothiol biosynthetic pathway that involves the deacetylation of 1-O-(2-acetamido-2-deoxy-α-D-glucopyranosyl)-D-myo-inositol (GlcNAc-Ins), via cleavage of an amide bond, to 1-O-(2-amino-2-deoxy-α-D-glucopyranosyl)-D-myo-inositol (GlcN-Ins) and acetate. In this study, MshB was expressed, purified and crystallized. A

Articles

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Questions

1–3 of 3 Questions  
  1. Are there any special storage conditions that must be maintained?

    1 answer
    1. No, Redi-Dri™ products can be stored under the same conditions as the equivalent non-Redi-Dri™ products.

      Helpful?

  2. How long will material remain free-flowing after it is opened?

    1 answer
    1. While the exact amount of time your material will remain free-flowing can vary, this largely depends on the ambient storage conditions and how many times it is opened.

      Helpful?

  3. How does Redi-Dri™ help keep salts and buffers free-flowing?

    1 answer
    1. Redi-Dri™ products are specially handled and the bottle is specially designed to ensure that the product will remain dry without adding impurities or additives to the product. Ensuring that you receive the highest quality material that is easy to handle.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service