Skip to Content
Merck

A4418

dd_molecule_design

Molecular Biology, ≥99.0%

Synonym(s):

Mascagnite, Ammonium sulfate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
(NH4)2SO4
CAS Number:
Molecular Weight:
132.14
PubChem Substance ID:
eCl@ss:
38030408
UNSPSC Code:
12352314
NACRES:
NA.31
EC Number:
231-984-1
MDL number:
Assay:
≥99.0%
Grade:
Molecular Biology
Form:
powder
Solubility:
H2O: 0.667 g/mL
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

Molecular Biology

Quality Level

assay

≥99.0%

form

powder

technique(s)

affinity binding assay: suitable, protein purification: suitable

impurities

≤0.005% Insolubles

pH

5.0-6 (25 °C, 132 g/L)

mp

>280 °C (dec.) (lit.)

solubility

H2O: 0.667 g/mL

density

1.77 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤5 ppm

cation traces

Cu: ≤5 ppm, Fe: ≤5 ppm, Zn: ≤5 ppm, heavy metals (as Pb): ≤2 ppm

absorption

≤0.025 at 280 at 1 M, ≤0.03 at 260 at 1 M

suitability

suitable for flow cytometry, suitable for molecular biology

application(s)

advanced drug delivery
metabolomics
microbiology

foreign activity

DNase, RNase, and protease, none detected

SMILES string

N.N.OS(O)(=O)=O

InChI

1S/2H3N.H2O4S/c;;1-5(2,3)4/h2*1H3;(H2,1,2,3,4)

InChI key

BFNBIHQBYMNNAN-UHFFFAOYSA-N

General description

Ammonium sulfate is a versatile inorganic sulfate salt, plays crucial role in biochemical research, molecular biology, and cell culture. It serves as an indispensable tool for purifying antibodies and precipitating or fractionating proteins. Furthermore, it is instrumental in crystallographic analysis of nucleic acids and proteins and also used as a nitrogen supplement in synthetic laboratory media. Our premium-grade Ammonium Sulfate is meticulously crafted to be DNase, RNase, and Protease-free, ensuring its suitability for cutting-edge research.

Application

Ammonium sulfate helps in quick precipitation of celluar proteins. Ammonium sulfate may be used for the precipitation or fractionation of proteins or for purification of antibodies. It is also useful for crystallographic analysis of nucleic acids and proteins.Ammonium sulfate has been used
  • in the preparation of ′Blue Silver′ colloidal Coomassie stain for proteins.
  • to precipitate the Shiga-like toxin from the culture supernatant.
  • for precipitaion of viral particles from the culture supernatant.

Features and Benefits

  • Heavy metals (as Pb): ≤2 ppm
  • Free from DNase, RNase, and protease

Other Notes

For additional information on our range of Biochemicals, please complete this form.


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content

Product Information Sheet


Assessment of Cell Cycle and Induction of Apoptosis by Shiga-like Toxin Produced by Escherichia coli O157:H7 in T47D Breast Cancer Cells Using Flow Cytometry
Wayan Suardana et al.
Asian Pacific Journal of Cancer Prevention, 23-10, 3247-3252 (2022)
Use of ammonium sulphate as a sulphur fertilizer: Implications for ammonia volatilization
David S Powlson, Chris J Dawson
Soil use and management, 38(1), 622-634 (2022)
Protein precipitation using ammonium sulfate
P Wingfield
Current Protocols in Protein Science / Editorial Board, John E. Coligan ... [Et Al.], 3 (2001)



Global Trade Item Number

SKUGTIN
A4418-5KG04061833374474
A4418-100G04061835542093
A4418-1KG04061835542109
A4418-500G04061835542116