Skip to Content
Merck
All Photos(1)

Key Documents

278424

Sigma-Aldrich

Poly(vinylsulfonic acid, sodium salt) solution

30-40 wt. % in H2O, technical grade

Synonym(s):

PVSA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H3NaO3S)n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

grade

technical grade

form

liquid

concentration

30-40 wt. % in H2O

refractive index

n20/D 1.389

density

1.267 g/mL at 25 °C

SMILES string

[Na]OS(=O)(=O)C=C

InChI

1S/C2H4O3S.Na/c1-2-6(3,4)5;/h2H,1H2,(H,3,4,5);/q;+1/p-1

InChI key

BWYYYTVSBPRQCN-UHFFFAOYSA-M

Application

Poly(vinylsulfonicacid, sodium salt) (PVSA) can be used:

  • In the preparation of superabsorbent semi-IPN (interpenetrating polymer network) hydrogel.
  • As a solid electrolyte for proton conduction in the fabrication of an all-solid-supercapacitor.
  • As a crystallization controlling agent in the preparation of high-quality crystals of porous coordination polymers(CP). PVSA regulates not only the size and structure of the crystals but also their preference orientation, resulting in CP channel alignment in the bulk powdery state.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shaoling Cheng et al.
Journal of chromatography. A, 1108(1), 43-49 (2006-01-31)
The goal of this work was to investigate the synergistic effect between the electrostatic and hydrophobic interactions upon the uptake of organic ions with hydrophobic moieties by ion-exchange resins with hydrophobic matrixes. The uptake of neutral amino acids by a
Ozlem Colak et al.
Artificial cells, blood substitutes, and immobilization biotechnology, 40(5), 354-361 (2012-05-01)
In this study, a novel amperometric glucose biosensor was developed by immobilizing glucose oxidase (GOX) by cross-linking via glutaraldehyde on electrochemically polymerized polypyrrole-poly(vinyl sulphonate) (PPy-PVS) films on the surface of a platinum (Pt) electrode. Electropolymerization of pyrrole and poly(vinyl sulphonate)
Srivatsan Kidambi et al.
Journal of the American Chemical Society, 126(50), 16286-16287 (2004-12-17)
The development of new methods for fabricating thin films that provide precise control of the three-dimensional topography and cell adhesion could lead to significant advances in the fields of tissue engineering and biosensors. This Communication describes the successful attachment and
Fatma Arslan et al.
Sensors (Basel, Switzerland), 11(8), 8152-8163 (2011-12-14)
In this study, a novel amperometric glucose biosensor with immobilization of glucose oxidase on electrochemically polymerized polyaniline-polyvinylsulphonate (Pani-Pvs) films has been accomplished via the entrapment technique. Electropolymerization of aniline on the Pt surface of the Pt electrode was carried out
Jitka Caslavska et al.
Electrophoresis, 27(23), 4618-4630 (2006-12-01)
Bidirectional ITP in fused-silica capillaries double-coated with Polybrene and poly-(vinylsulfonate) is a robust approach for analysis of low-molecular-mass compounds. EOF towards the cathode is strong (mobility >4.0 x 10(-8) m(2)/Vs) within the entire pH range investigated (2.40-8.08), dependent on ionic

Articles

Recently, layer-by-layer (LbL) assembly has emerged as a versatile, gentle and, simple method for immobilization of functional molecules in an easily controllable thin film morphology.1,2 In this short review, we introduce recent advances in functional systems fabricated by using the mild, yet adaptable LbL technique.

We present an article that discusses two applications in particular; first, using these layers as polyelectrolyte membranes to control permeability.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service