Skip to Content
Merck
All Photos(1)

Key Documents

80154

Supelco

Dipentyl phthalate

Selectophore, ≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H26O4
CAS Number:
Molecular Weight:
306.40
Beilstein:
1987323
EC Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

grade

for ion-selective electrodes

Quality Level

product line

Selectophore

Assay

≥99.0% (GC)
≥99.0%

form

liquid

refractive index

n20/D 1.490

density

1.025 g/mL at 20 °C (lit.)

SMILES string

CCCCCOC(=O)c1ccccc1C(=O)OCCCCC

InChI

1S/C18H26O4/c1-3-5-9-13-21-17(19)15-11-7-8-12-16(15)18(20)22-14-10-6-4-2/h7-8,11-12H,3-6,9-10,13-14H2,1-2H3

InChI key

IPKKHRVROFYTEK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dipentyl phthalate (DPeP) belongs to phthalate group, and is a dialkyl or aryl/alkyl diesters of phthalic acid. It may be prepared from phthalic anhydride along with n-pentanol.
Visit our Sensor Applications portal to learn more.

Application


  • Biologically relevant reductions in fetal testosterone and Insl3 induced by in utero exposure to high levels of di-isononyl phthalate (DINP) in male rats: Dipentyl phthalate′s structural similarities to DINP underpin studies on phthalate exposure′s impacts on male reproductive health, providing insights into potential risks and mechanisms of toxicity in developmental stages (Earl Gray L Jr, 2023).

  • Exposure to dipentyl phthalate in utero disrupts the adrenal cortex function of adult male rats by inhibiting SIRT1/PGC-1α and inducing AMPK phosphorylation: This study uses dipentyl phthalate to examine the adverse effects of phthalate exposure on adrenal gland function, contributing to a deeper understanding of metabolic disruptions (Chen et al., 2023).

  • Determination of 15 phthalic acid esters based on GC-MS/MS coupled with modified QuEChERS in edible oils: This research demonstrates the utility of dipentyl phthalate in method development for detecting phthalates in food matrices, enhancing food safety assessments (Wang et al., 2022).

  • Development of a putative adverse outcome pathway network for male rat reproductive tract abnormalities with specific considerations for the androgen sensitive window of development: Dipentyl phthalate is investigated for its role in developmental toxicity, aiding in the construction of models predicting reproductive health effects from chemical exposures (Palermo et al., 2021).

Other Notes

Plasticizer for PVC membrane electrodes

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

244.4 °F - closed cup

Flash Point(C)

118 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M.M. Khalil
Analytical Letters, 26, 2071-2071 (1993)
J J Heindel et al.
Toxicology and applied pharmacology, 97(2), 377-385 (1989-02-01)
High doses of phthalate esters in vivo cause testicular lesions. One initial target for these effects is the sustentacular Sertoli cell. Sertoli cells are unique in that they have surface membrane receptors for follicle-stimulating hormone (FSH), which couple to adenylate
D M Creasy et al.
Experimental and molecular pathology, 46(3), 357-371 (1987-06-01)
A sequential morphological study has been carried out to examine the ultrastructural effects of di-n-pentyl phthalate (DPP) on the mature rat testis. A single oral dose of 2.2 g DPP/kg body wt was administered, and testes, perfuse-fixed 3-48 hr after
N P Moore et al.
Archives of toxicology, 66(6), 435-442 (1992-01-01)
Cell-specific testicular toxicants have been used to examine the distribution of creatine within the rat testis, and to examine the potential use of creatinuria as a non-invasive indicator of testicular damage. Groups of rats were administered single doses of various
Changes in ultrastructure and cytochemical localization of zinc in rat testis following the administration of di-n-pentyl phthalate.
P M Foster et al.
Toxicology and applied pharmacology, 63(1), 120-132 (1982-03-30)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service