Skip to Content
Merck

36934

Bis(methylglycol) phthalate

analytical standard

Synonym(s):

Bis(2-methoxyethyl) phthalate, Dimethylglycol phthalate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C14H18O6
CAS Number:
Molecular Weight:
282.29
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
EC Number:
204-212-6
Beilstein/REAXYS Number:
2056929
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

bp

230 °C/10 mmHg (lit.)

density

1.173 g/mL at 20 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

COCCOC(=O)c1ccccc1C(=O)OCCOC

InChI

1S/C14H18O6/c1-17-7-9-19-13(15)11-5-3-4-6-12(11)14(16)20-10-8-18-2/h3-6H,7-10H2,1-2H3

InChI key

HSUIVCLOAAJSRE-UHFFFAOYSA-N

General description

Bis(methylglycol) phthalate belongs to the class of phthalate esters widely used as plasticizers in plastic films.
This compound is listed in the SVHC (Substances of very high concern) candidate list of ECHA (European Chemicals Agency)

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support. Bis(methylglycol) phthalate may be used as an analytical reference standard for the quantification of the analyte in plastic food packaging materials using gas chromatography coupled to tandem mass spectrometry.

Other Notes

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.


Still not finding the right product?

Explore all of our products under Bis(methylglycol) phthalate


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Repr. 1B

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 1

flash_point_f

249.8 °F - closed cup

flash_point_c

121 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Gas chromatography-triple quadrupole tandem mass spectrometry for successive single-surface migration study of phthalate esters from polythene film
Yang J, et al.
Food Control, 73, 1134-1143 (2017)
Wataru Naito et al.
Environmental monitoring and assessment, 115(1-3), 451-471 (2006-05-02)
Screening-level ecological risk assessments of di(2-ethylhexyl)phthalate (DEHP) for aquatic organisms in Japan were conducted using estimated statistical values based on surface water and sediment monitoring data and effect threshold values based on a large aquatic toxicity database. An alternative method
J Campbell et al.
Journal of applied toxicology : JAT, 4(1), 35-41 (1984-02-01)
The rat foetus, in contrast with the maternal liver and placenta, has little or no ability to hydrolyse di-(2-methoxyethyl)-phthalate (DMEP) to mono-2-methoxyethyl)-phthalate (MMEP). At short times after the administration of DMEP to the dam on the 14th day of gestation



Global Trade Item Number

SKUGTIN
36934-250MG04061838185976