Skip to Content
Merck
All Photos(1)

Documents

79245

Supelco

(+)-Carvone

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

(+)-p-Mentha-6,8-diene 2-one, (S)-5-Isopropenyl-2-methyl-2-cyclohexenone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14O
CAS Number:
Molecular Weight:
150.22
Beilstein:
2042970
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

228-230 °C (lit.)

density

0.960 g/mL at 20 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

−20°C

SMILES string

CC(=C)[C@H]1CC=C(C)C(=O)C1

InChI

1S/C10H14O/c1-7(2)9-5-4-8(3)10(11)6-9/h4,9H,1,5-6H2,2-3H3/t9-/m0/s1

InChI key

ULDHMXUKGWMISQ-VIFPVBQESA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

This compound is commonly found in plants of the genus: carum mentha zingiber

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1A

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

204.1 °F - closed cup

Flash Point(C)

95.6 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

W Jäger et al.
The Journal of pharmacy and pharmacology, 53(5), 637-642 (2001-05-24)
The purpose of this study was to determine whether an enantioselective difference in the metabolism of topically applied R-(-)- and S-(+)-carvone could be observed in man. In a previous investigation we found that R-(-)- and S-(+)-carvone are stereoselectively biotransformed by
Raul Conde et al.
Phytomedicine : international journal of phytotherapy and phytopharmacology, 18(14), 1197-1201 (2011-08-02)
There is no universally accepted and effective prophylaxis of migraine headache episodes. Thus we aimed to investigate the effects of Lippia alba (Mill.) N. E. Brown, an herb with many effects on central nervous system, on pain frequency and intensity
Elissavet E Anagnostaki et al.
Organic letters, 15(1), 152-155 (2012-12-22)
The synthesis of hydroxyelemane 5 from (R)-carvone and its utilization as a common synthetic scaffold to produce structurally diverse germacrane and guaiane sesquiterpenes are described. A highly enantio- and stereoselective biomimetic tandem oxy-Cope/ene rearrangement was used as the key reaction
V Y Hatano et al.
Brazilian journal of medical and biological research = Revista brasileira de pesquisas medicas e biologicas, 45(3), 238-243 (2012-02-24)
Lippia alba (Mill.) N.E. Brown (Verbenaceae) is widely used in different regions of Central and South America as a tranquilizer. The plant's anxiolytic properties, however, merit investigation. The present study evaluated the effects of repeated daily (14 days) intraperitoneal (ip)
Marta Goretti et al.
Bioresource technology, 121, 290-297 (2012-08-04)
Response surface methodology was applied in optimizing the asymmetric bioreduction of (4S)-(+)-carvone to dihydrocarvone (with low incidence of unsought side reactions) by using whole-cells of Cryptococcus gastricus. A factorial design (2(5)) including five independent variables was performed: X(1)=incubation time; X(2)=pH;

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service