Skip to Content
Merck
All Photos(1)

Key Documents

W224928

Sigma-Aldrich

D-Carvone

≥96%, FG

Synonym(s):

(S)-(+)-Carvone, (+)-Carvone, (S)-5-Isopropenyl-2-methyl-2-cyclohexenone, p-Mentha-6,8-dien-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14O
CAS Number:
Molecular Weight:
150.22
FEMA Number:
2249
Beilstein:
2042970
EC Number:
Council of Europe no.:
146
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
7.146
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

Assay

≥96%

composition

contains IFRA restricted Carvone

refractive index

n20/D 1.497 (lit.)

bp

96-98 °C/10 mmHg (lit.)

density

0.96 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

carvone

Organoleptic

caraway; minty

SMILES string

CC(=C)[C@H]1CC=C(C)C(=O)C1

InChI

1S/C10H14O/c1-7(2)9-5-4-8(3)10(11)6-9/h4,9H,1,5-6H2,2-3H3/t9-/m0/s1

InChI key

ULDHMXUKGWMISQ-VIFPVBQESA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

D-Carvone is a monoterpene ketone that can be used as a flavoring agent. It occurs naturally in essential oil of caraway seeds and citrus fruits.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1A

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

204.1 °F - closed cup

Flash Point(C)

95.6 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Burdock GA
Encyclopedia of Food and Color Additives, 1, 522-523 (1997)
Cancer chemoprevention by phytochemicals in fruits and vegetables: an overview.
Huang MT, et al
ACS Symp. Ser., 546, 2?16-2?16 (1994)
Beneficial effect of carvone, a dietary monoterpene ameliorates hyperglycemia by regulating the key enzymes activities of carbohydrate metabolism in streptozotocin-induced diabetic rats
Muruganathan U & Srinivasan S.
Biomedicine and Pharmacotherapy, 84, 1558-1567 (2016)
Determination of enantiomer ratios of d, l-carvone in supercritical fluid extracts from caraway seeds and speamint leaves by high-performance liquid chromatography with polarimetric and ultraviolet spectrophotometric detection
Bounoshita M, et al
Analytical Sciences, 9(3), 425-428 (1993)
Mengyang Xuan et al.
Organic letters, 14(21), 5492-5495 (2012-10-26)
The total synthesis of the cAMP signaling pathway activator (-)-alotaketal A is reported. A convergent approach to the unusual alotane sesterterpenoid skeleton was employed, exploiting a remarkable LiDBB-mediated coupling of an (R)-carvone-derived δ-lactone with an allyl bromide side chain, followed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service