Skip to Content
Merck
All Photos(1)

Documents

76524

Supelco

Glycine

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

Aminoacetic acid, Aminoethanoic acid, Glycocoll

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2COOH
CAS Number:
Molecular Weight:
75.07
Beilstein:
635782
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
E Number:
E640
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

analyte chemical class(es)

amino acids, peptides, proteins

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

color

white

pKa (25 °C)

(1) 2.35, (2) 9.60
2.35

mp

240 °C (dec.) (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

20-25°C

SMILES string

NCC(O)=O

InChI

1S/C2H5NO2/c3-1-2(4)5/h1,3H2,(H,4,5)

InChI key

DHMQDGOQFOQNFH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Check out the entire portfolio of our amino acids certified reference materials (CRM)

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Inhibitory neurotransmitter in spinal cord, allosteric regulator of NMDA receptors.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexander Carvajal-González et al.
Brain : a journal of neurology, 137(Pt 8), 2178-2192 (2014-06-22)
The clinical associations of glycine receptor antibodies have not yet been described fully. We identified prospectively 52 antibody-positive patients and collated their clinical features, investigations and immunotherapy responses. Serum glycine receptor antibody endpoint titres ranged from 1:20 to 1:60 000.
Li-Chun Tu et al.
The EMBO journal, 32(24), 3220-3230 (2013-11-12)
Nuclear pore complexes (NPCs) mediate cargo traffic between the nucleus and the cytoplasm of eukaryotic cells. Nuclear transport receptors (NTRs) carry cargos through NPCs by transiently binding to phenylalanine-glycine (FG) repeats on intrinsically disordered polypeptides decorating the NPCs. Major impediments
Zoé Husson et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 34(28), 9418-9431 (2014-07-11)
The principal neurons of the cerebellar nuclei (CN), the sole output of the olivo-cerebellar system, receive a massive inhibitory input from Purkinje cells (PCs) of the cerebellar cortex. Morphological evidence suggests that CN principal cells are also contacted by inhibitory
Suzanne L Grant et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 844(2), 278-282 (2006-08-08)
A simple and versatile methodology using high-performance liquid chromatography (HPLC) with fluorimetric detection was developed to simultaneously determine d-serine along with other metabolically related neuroactive amino acids in the glutamatergic system: L-serine, L-glutamate, L-glutamine, and glycine. On-column sensitivity was in
Steven Tran et al.
Behavioural brain research, 276, 161-170 (2014-03-07)
The zebrafish is emerging as a popular animal model for alcohol (ethanol or EtOH) addiction due to its simplicity and practical advantages. Two phenomena associated with ethanol addiction are the development of tolerance and withdrawal. Using a multi-level approach in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service