Merck
  • An integrative analysis of ethanol tolerance and withdrawal in zebrafish (Danio rerio).
An integrative analysis of ethanol tolerance and withdrawal in zebrafish (Danio rerio).
Behavioural brain research (2014-03-07)
Steven Tran, Diptendu Chatterjee, Robert Gerlai
ABSTRACT
The zebrafish is emerging as a popular animal model for alcohol (ethanol or EtOH) addiction due to its simplicity and practical advantages. Two phenomena associated with ethanol addiction are the development of tolerance and withdrawal. Using a multi-level approach in the current study, we characterise ethanol tolerance and withdrawal in zebrafish. We first investigate the temporal trajectory of ethanol concentration in the zebrafish brain in response to an acute exposure and during withdrawal. We report that ethanol concentrations approach a steady state within 60 min of exposure to 0.50% and 1.00% v/v ethanol and rapidly decline and return to zero within 60 min following withdrawal from chronic ethanol exposure (0.50% v/v). We characterise the changes associated with ethanol tolerance and withdrawal in zebrafish by focusing on three domains relevant to ethanol addiction: motor patterns, physiological responses (i.e. cortisol levels), and neurochemical alterations. The use of multiple domains of investigation allowed an in-depth analysis of ethanol induced changes in zebrafish.
Sigma-Aldrich
Taurine, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Taurine, suitable for cell culture, meets USP testing specifications
Sigma-Aldrich
Taurine, ≥99%
Supelco
Taurine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Taurine, ≥98%, FG
SAFC
Taurine
USP
Taurine, United States Pharmacopeia (USP) Reference Standard
Hydrocortisone for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Glycine, analytical standard, for nitrogen determination according to Kjeldahl method
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Glycine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
3,4-Dihydroxyphenylacetic acid, 98%
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
SAFC
Glycine
Sigma-Aldrich
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Glycine, tested according to Ph. Eur.
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Hydrocortisone, British Pharmacopoeia (BP) Assay Standard
Supelco
3,4-Dihydroxyphenylacetic acid, analytical standard