Skip to Content
Merck
All Photos(1)

Documents

33226

Sigma-Aldrich

Glycine

puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)

Synonym(s):

Aminoacetic acid, Aminoethanoic acid, Glycocoll

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2COOH
CAS Number:
Molecular Weight:
75.07
Beilstein:
635782
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.21

Agency

USP/NF
reag. Ph. Eur.

Quality Level

grade

puriss. p.a.

Assay

99.7-101% (calc. to the dried substance)

form

crystalline powder

technique(s)

IR spectroscopy: suitable

impurities

≤0.001% heavy metals (as Pb)
≤0.02% ammonium (NH4)
≤0.1% water (Karl Fischer)
≤1.0% ninhydrin-positive substances

ign. residue

≤0.05% (as SO4)

loss

≤0.5% loss on drying, 105°C, 2 h

pH

5.9-6.4 (20 °C, 5%)

pKa (25 °C)

(1) 2.35, (2) 9.60
2.35

mp

240 °C (dec.) (lit.)

anion traces

chloride (Cl-): ≤30 mg/kg
sulfate (SO42-): ≤20 mg/kg

cation traces

Fe: ≤5 mg/kg

suitability

in accordance for identity (IR)

SMILES string

NCC(O)=O

InChI

1S/C2H5NO2/c3-1-2(4)5/h1,3H2,(H,4,5)

InChI key

DHMQDGOQFOQNFH-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Glycine can be used as:
  • A reactant in the natural soil clay-catalyzed cleavage and polycondensation with pyrogallol.
  • A catalyst in the Knoevenagel reaction between aryl aldehydes and malononitrile to form a C=C bond.
  • A reagent in the metal-free synthesis of biaryls from aryl sulfoxides and sulfonanilides via sigmatropic rearrangement.

Biochem/physiol Actions

Inhibitory neurotransmitter in spinal cord, allosteric regulator of NMDA receptors.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Richard A Felix et al.
Brain structure & function, 220(5), 2639-2652 (2014-06-30)
The mammalian superior paraolivary nucleus (SPON) is a major source of GABAergic inhibition to neurons in the inferior colliculus (IC), a well-studied midbrain nucleus that is the site of convergence and integration for the majority ascending auditory pathways en route
Alexander Carvajal-González et al.
Brain : a journal of neurology, 137(Pt 8), 2178-2192 (2014-06-22)
The clinical associations of glycine receptor antibodies have not yet been described fully. We identified prospectively 52 antibody-positive patients and collated their clinical features, investigations and immunotherapy responses. Serum glycine receptor antibody endpoint titres ranged from 1:20 to 1:60 000.
Li-Chun Tu et al.
The EMBO journal, 32(24), 3220-3230 (2013-11-12)
Nuclear pore complexes (NPCs) mediate cargo traffic between the nucleus and the cytoplasm of eukaryotic cells. Nuclear transport receptors (NTRs) carry cargos through NPCs by transiently binding to phenylalanine-glycine (FG) repeats on intrinsically disordered polypeptides decorating the NPCs. Major impediments
Zoé Husson et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 34(28), 9418-9431 (2014-07-11)
The principal neurons of the cerebellar nuclei (CN), the sole output of the olivo-cerebellar system, receive a massive inhibitory input from Purkinje cells (PCs) of the cerebellar cortex. Morphological evidence suggests that CN principal cells are also contacted by inhibitory
Suzanne L Grant et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 844(2), 278-282 (2006-08-08)
A simple and versatile methodology using high-performance liquid chromatography (HPLC) with fluorimetric detection was developed to simultaneously determine d-serine along with other metabolically related neuroactive amino acids in the glutamatergic system: L-serine, L-glutamate, L-glutamine, and glycine. On-column sensitivity was in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service