Skip to Content
Merck
All Photos(1)

Key Documents

76235

Supelco

Paraffin oil

suitable for IR spectroscopy

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12000000
NACRES:
NA.21
technique(s):
IR spectroscopy: suitable
bp:
300 °C (lit.)
Pricing and availability is not currently available.

form

oil

Quality Level

technique(s)

IR spectroscopy: suitable

refractive index

n20/D 1.473

viscosity

100-145 mPa.s(20 °C)

bp

300 °C (lit.)

mp

-24 °C (lit.)

Looking for similar products? Visit Product Comparison Guide

General description

Paraffin oil resists water. It is widely used as release agent, lubricant, defoamer and also as coating component.

Application

Paraffin oil has been used in preparation of tyrosinase-modified carbon paste electrode for detection of Catechol.[1]

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

419.0 °F - closed cup

Flash Point(C)

215 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Eryn Slankster et al.
Scientific reports, 10(1), 3119-3119 (2020-02-23)
Starvation enhances olfactory sensitivity that encourage animals to search for food. The molecular mechanisms that enable sensory neurons to remain flexible and adapt to a particular internal state remain poorly understood. Here, we study the roles of GABA and insulin
Rate-limiting steps of tyrosinase-modified electrodes for the detection of catechol.
Burestedt E
Analytical Chemistry, 68(9), 1605-1611 (1996)
Wenming Wu et al.
The Analyst, 136(11), 2287-2293 (2011-04-05)
Bubble formation inside a microscale channel is a significant problem in general microfluidic experiments. The problem becomes especially crucial when performing a polymerase chain reaction (PCR) on a chip which is subject to repetitive temperature changes. In this paper, we
Yi-Shiung Horng et al.
American journal of physical medicine & rehabilitation, 90(6), 435-442 (2011-03-25)
: The aim of this study was to investigate the effectiveness of tendon and nerve gliding exercises as a part of combined treatments for carpal tunnel syndrome. : Patients with carpal tunnel syndrome were randomized into three groups. All patients
Jeryong Kim et al.
Surgery today, 41(12), 1680-1683 (2011-10-05)
Paraffin oil injection into the breast, which had been used for breast augmentation in the past, can cause various complications. Complete removal of paraffinoma lesions with immediate breast reconstruction using autogenous tissue is a very satisfactory treatment option. However, diffuse

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service