Skip to Content
Merck
All Photos(1)

Documents

16415

Sigma-Aldrich

Vaseline®

meets analytical specification of Ph. Eur.

Synonym(s):

Petrolatum

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
53131641
NACRES:
NA.21

description

liquefying point 35 - 70 °C

Quality Level

quality

meets analytical specification of Ph. Eur.

technique(s)

IR spectroscopy: suitable

impurities

acidity or alcalinity, in accordance
residual solvents, in accordance
≤300 ppm aromatic polycycl. hydrocarbons

ign. residue

≤0.05% (as SO4)

solubility

water: insoluble

suitability

in accordance for identity (IR)
passes test for appearance of solution

application(s)

pharmaceutical (small molecule)

Looking for similar products? Visit Product Comparison Guide

Application


  • Strychnos pseudoquina modulates the morphological reorganization of the scar tissue of second intention cutaneous wounds in rats.: This study examines the use of Vaseline in combination with Strychnos pseudoquina to enhance scar tissue healing and reorganization in cutaneous wounds. The findings highlight the biochemical pathways involved in wound healing and the potential therapeutic applications of Vaseline in dermatological treatments (Sarandy et al., 2018).

  • Schinus terebinthifolius Raddi (Aroeira) leaves oil attenuates inflammatory responses in cutaneous wound healing in mice.: The application of Vaseline as a base for delivering Aroeira leaves oil demonstrates significant anti-inflammatory effects in cutaneous wound healing. This research underscores the biochemical mechanisms through which Vaseline enhances the therapeutic properties of herbal extracts in wound care (Estevao et al., 2017).

  • Liquid Crystal Formation from Sunflower Oil: Long Term Stability Studies.: The study explores the formulation of stable liquid crystals from sunflower oil using Vaseline as a stabilizing agent. The findings highlight the molecular interactions and long-term stability of such formulations, emphasizing their potential applications in biochemistry and material science (da Rocha-Filho et al., 2016).

Legal Information

Vaseline is a registered trademark of Unilever Supply Chain, Inc.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tomomi Tani et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 25(9), 2181-2191 (2005-03-05)
The behavior of single molecules of neurotrophins on growth cones was observed by the use of the fluorescent conjugate of nerve growth factor (NGF), Cy3-NGF. After the application of 0.4 nm Cy3-NGF, chick dorsal root ganglion growth cones responded within
Lucie Jelínková et al.
Biology of reproduction, 74(5), 905-912 (2006-02-03)
Aurora kinase B (AURKB) is a chromosomal passenger protein that is essential for a number of processes during mitosis. Its activity is regulated by association with two other passenger proteins, INCENP and Survivin, and by phosphorylation on Thr 232. In
Mehrnaz Geranmayeh et al.
Archives of gynecology and obstetrics, 285(1), 77-81 (2011-05-27)
Maintaining an intact perineum is a highly regarded aim in delivery procedures today. Since perineal massage is a common practice during delivery, the present study aims to investigate the effect of perineal massage with Vaseline on perineal trauma (rate of
Jeremiah A Alt et al.
American journal of otolaryngology, 33(4), 469-472 (2011-12-14)
We sought to determine the efficacy of commonly used earplugs using an anatomically correct ear model. The total volume and rate of water that leaked past the earplug and subsequent defect in the tympanic membrane over separately measured 30, 60
Ágnes Rosecker et al.
The journal of sexual medicine, 10(2), 509-515 (2012-08-29)
Vaseline self-injection into the penis is currently a popular procedure in prisons. Since such injections are illegal, severe complications of the procedure can often remain hidden. To identify the incidence, motivation, and morbidity of Vaseline self-injection into the penis among

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service