Skip to Content
Merck
All Photos(1)

Key Documents

75160

Supelco

Methyl oleate

Methyl oleate

analytical standard

Synonym(s):

Methyl cis-9-octadecenoate, Oleic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7CH=CH(CH2)7CO2CH3
CAS Number:
Molecular Weight:
296.49
Beilstein:
1727037
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

vapor pressure

10 mmHg ( 205 °C)

Assay

≥98.5% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.452 (lit.)
n20/D 1.452

bp

218 °C/20 mmHg (lit.)

density

0.874 g/mL at 20 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

functional group

ester

shipped in

ambient

storage temp.

2-8°C

SMILES string

CCCCCCCC\C=C/CCCCCCCC(=O)OC

InChI

1S/C19H36O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19(20)21-2/h10-11H,3-9,12-18H2,1-2H3/b11-10-

InChI key

QYDYPVFESGNLHU-KHPPLWFESA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl oleate is a methyl ester of the monounsaturated fatty acid (MUFA), oleic acid, which is abundantly found in dietary fats and oils.

Application

This analytical standard can also be used as follows:

  • Determination of free fatty acids in plasma samples following their methylation by gas chromatography (GC)
  • Comparative analysis of gas chromatography-combustion-mass spectrometry and gas chromatography-flame ionization detector methods for the determination of fatty acid methyl esters (FAMEs) in biodiesel samples
  • Simultaneous determination of fatty acid methyl esters in commercial food oil samples by gas chromatography-vacuum ultraviolet (GC-VUV) spectroscopy
  • Measurement of fatty acid methyl ester composition of various edible oil samples by 1H nuclear magnetic resonance (1H NMR) spectroscopy combined with partial least squares (PLS) method
  • Simultaneous determination of fatty acids in bovine colostrum samples by GC-FID after their derivatization to ester forms using an acidic catalyst boron trifluoride

Other Notes

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Selective transesterification of triolein with methanol to methyl oleate and glycerol using alumina loaded with alkali metal salt as a solid-base catalyst
Ebiura T, et al.
Applied Catalysis A: General, 283(1), 111-116 (2005)
Selective hydrogenation of methyl oleate into unsaturated alcohols: Relationships between catalytic properties and composition of cobalt?tin catalysts
Pouilloux.Y, et al.
Catalysis Today, 63(1), 87-100 (2000)
Annapurna Kumari et al.
Biotechnology for biofuels, 2(1), 1-1 (2009-01-16)
Transesterification of Jatropha oil was carried out in t-butanol solvent using immobilized lipase from Enterobacter aerogenes. The presence of t-butanol significantly reduced the negative effects caused by both methanol and glycerol. The effects of various reaction parameters on transesterification of
Ursula Biermann et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 18(26), 8201-8207 (2012-05-18)
A kinetic study of the dodecanethiol-catalyzed cis/trans isomerization of methyl oleate (cis-2) without added initiator was performed by focusing on the initiation of the radical chain reaction. The reaction orders of the rate of isomerization were 2 and 0.5 for
Farshad Darvishi et al.
New biotechnology, 28(6), 756-760 (2011-02-18)
The yeast Yarrowia lipolytica degrades efficiently low-cost hydrophobic substrates for the production of various added-value products such as lipases. To obtain yeast strains producing high levels of extracellular lipase, Y. lipolytica DSM3286 was subjected to mutation using ethyl methanesulfonate (EMS)

Protocols

HPLC Analysis of Fatty Acid Methyl Esters (FAMES) on SUPELCOSIL™ LC-18

Questions

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service