Skip to Content
Merck
All Photos(1)

Key Documents

295329

Sigma-Aldrich

Ethylene

≥99.5%

Synonym(s):

Ethene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CH2
CAS Number:
Molecular Weight:
28.05
Beilstein:
1730731
EC Number:
MDL number:
UNSPSC Code:
12142100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

vapor density

0.97 (vs air)

Quality Level

vapor pressure

35.04 atm ( 20 °C)

Assay

≥99.5%

autoignition temp.

842 °F

expl. lim.

36 %

bp

−104 °C (lit.)

mp

−169 °C (lit.)

SMILES string

C=C

InChI

1S/C2H4/c1-2/h1-2H2

InChI key

VGGSQFUCUMXWEO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethylene is an alkene used in Friedel-Crafts alkylation, cycloaddition, electrophilic addition, and vinylation reactions.[1]

Application

Ethylene is mainly used as a precursor to synthesize polyethylene, ethylbenzene and ethylene dichloride.[2][3][4][5] It is also used in the preparation of single- layer graphene by thermal chemical vapor deposition.[6]

Packaging

Supplied in a carbon steel lecture bottle with a CGA180M/CGA110F needle valve installed.

Compatible with the following:
  • Aldrich® lecture-bottle station systems
  • Aldrich® lecture-bottle gas regulators

Other Notes

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC

hose barb

Product No.
Description
Pricing

purge valve

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Gas 1 - Press. Gas Liquefied gas - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

2A - Gases

WGK

nwg

Flash Point(F)

-148.0 °F - closed cup

Flash Point(C)

-100 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Addition of arenes to ethylene and propene catalyzed by ruthenium.
Lail, Marty and Arrowood, Benjamin N and Gunnoe, T Brent
Journal of the American Chemical Society, 125(25), 7506-7507 (2003)
Ethylene
Crimmins MT and Kim-Meade AS
eEROS (Encyclopedia of Reagents for Organic Synthesis) (2001)
Catalytic hydroarylation of ethylene using TpRu (L)(NCMe) Ph (L= 2, 6, 7-Trioxa-1-phosphabicyclo [2, 2, 1] heptane): Comparison to TpRu (L?)(NCMe) Ph systems (L?= CO, PMe3, P (pyr) 3, or P (OCH2) 3CEt).
Joslin, Evan E et al.
Organometallics, 31(19), 6851-6860 (2012)
Pyridinic N doped graphene: synthesis, electronic structure, and electrocatalytic property.
Luo, Zhiqiang et al.
Journal of Materials Chemistry, 21(22), 8038-8044 (2011)
A new catalyst for highly syndiospecific living olefin polymerization: homopolymers and block copolymers from ethylene and propylene.
Tian, Jun et al.
Journal of the American Chemical Society, 123(21), 5134-5135 (2001)

Protocols

Separation of Methane; Acetylene; Carbon monoxide; Water; Nitrogen; Carbon dioxide; Ethane; Ethylene

Questions

1–3 of 3 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. What type of regulator would work for the ethylene bottle?

    1 answer
    1. As per the related products section, Products Z146706 and Z512958 are the recommended regulators for Product 295329.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service