Skip to Content
Merck
All Photos(1)

Key Documents

N29406

Sigma-Aldrich

Nonane

ReagentPlus®, 99%

Synonym(s):

n-Nonane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7CH3
CAS Number:
Molecular Weight:
128.26
Beilstein:
1696917
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.21
grade:
reagent
bp:
151 °C (lit.)
vapor pressure:
0.18 psi ( 37.7 °C)

grade

reagent

Agency

suitable for EPA 1613

vapor density

4.41 (vs air)

vapor pressure

0.18 psi ( 37.7 °C)

product line

ReagentPlus®

Assay

99%

form

liquid

autoignition temp.

401 °F

expl. lim.

2.9 %

dilution

(for general lab use)

refractive index

n20/D 1.405 (lit.)

bp

151 °C (lit.)

mp

−53 °C (lit.)

density

0.718 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCC

InChI

1S/C9H20/c1-3-5-7-9-8-6-4-2/h3-9H2,1-2H3

InChI key

BKIMMITUMNQMOS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Nonane is a linear alkane that is mainly obtained from petroleum distillate. It is used as a solvent in organic synthesis.

Application

Nonane (n-Nonane) can be used as a solvent in:      
  • The Ullman reaction and the hydrogenation of CO2 to form CH3OH in the presence of a copper catalyst.      
  • The porosity characterization of different carbons by “nonane blocking of micropores”.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

87.8 °F - closed cup

Flash Point(C)

31.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Characterization of single wall carbon nanotubes by nonane preadsorption.
Byl O, et al.
Carbon, 44(10), 2039-2044 (2006)
Measurement of triglycerides using nonane extraction and colorimetry.
A L Levy
Annals of clinical laboratory science, 2(6), 474-479 (1972-11-01)
Oxidation kinetics of n-nonane: Measurements and modeling of ignition delay times and product concentrations.
Rotavera B, et al.
Proceedings of the Combustion Institute, 33(1), 175-183 (2011)
Heat capacities and densities of liquid n-octane, n-nonane, n-decane, and n-hexadecane at temperatures from 318.15 K to 373.15 K and at pressures up to 10 MPa.
Banipal TS, et al.
The Journal of Chemical Thermodynamics, 23(10), 923-931 (1991)
Lip Kim Lee et al.
Applied and environmental microbiology, 76(3), 794-802 (2009-12-17)
Polybrominated diphenyl ethers (PBDEs) have attracted attention recently due to their proven adverse effects on animals and their increasing concentrations in various environmental media and biota. To gain insight into the fate of PBDEs, microcosms established with soils and sediments

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service