Skip to Content
Merck
All Photos(1)

Key Documents

72240

Sigma-Aldrich

Aluminum-nickel alloy

purum, 50% Al basis, 50% Ni basis

Synonym(s):

Nickel-aluminum alloy, Raney®-Nickel alloy

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Quality Level

grade

purum

form

powder

composition

Al, 50%
Ni, 50%

reaction suitability

core: nickel
reagent type: catalyst

SMILES string

[Al].[Ni]

InChI

1S/Al.Ni

InChI key

NPXOKRUENSOPAO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Aluminum-nickel alloy can be used:
  • For the reduction of nitriles, nitro compounds and oximes to primary amines.
  • For the conversion of carbonyl compounds to hydrocarbons.
  • As a reducing agent to convert Ar-X derivatives to Ar-H compounds.
  • For the synthesis of Raney nickel catalyst to be used as an alkylating agent.
  • To synthesize non-pyrophoric Ni catalyst by alkali leaching.
  • For the synthesis of alloyed skeletal Ni catalysts.

Legal Information

Raney is a registered trademark of W.R. Grace & Co.-Conn.

replaced by

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Flam. Sol. 1 - Skin Sens. 1 - Water-react 2

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reduction of carbonyl groups to the corresponding methylenes with Ni-Al alloy in water.
Ishimoto K, et al.
Chemical Communications (Cambridge, England), 89(4), 514-515 (2003)
Highly selective hydrogenation of C?C double bond in unsaturated carbonyl compounds over NiC catalyst.
Mahata N, et al.
Chemical Engineering Journal, 188(4), 155-159 (2012)
An environmentally benign and catalytically efficient non-pyrophoric Ni catalyst for aqueous-phase reforming of ethylene glycol.
Zhu LJ, et al.
Green Chemistry, 10(12), 1323-1330 (2008)
Reinvestigating Raney nickel mediated selective alkylation of amines with alcohols via hydrogen autotransfer methodology.
Mehta A, et al.
Applied Catalysis A: General, 478(4), 241-251 (2014)
Steam reforming of ethanol over skeletal Ni-based catalysts: A temperature programmed desorption and kinetic study
Zhang C, et al.
AIChE Journal, 60(2), 635-644 (2014)

Articles

Mechanical alloying is a “brute force” method of affecting alloying and chemical reactions. The mixture of reactant powders and several balls are placed in the milling jar of a high-energy ball mill, for example, a shaker mill or a planetary mill.

Nickel complexes catalyze various synthetic reactions like oxidative addition, C-H activation, and cross-coupling.

Nickel complexes catalyze various synthetic reactions like oxidative addition, C-H activation, and cross-coupling.

Nickel complexes catalyze various synthetic reactions like oxidative addition, C-H activation, and cross-coupling.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service