Skip to Content
Merck
All Photos(1)

Key Documents

221678

Sigma-Aldrich

Raney®-Nickel

W.R. Grace and Co. Raney® 2800, slurry, in H2O, active catalyst

Synonym(s):

Raney Ni, Nickel sponge

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
AlNi
CAS Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Quality Level

form

slurry

composition

Ni, ≥89%

reaction suitability

core: nickel
reagent type: catalyst

manufacturer/tradename

W.R. Grace and Co. Raney® 2800

concentration

in H2O

impurities

6-9% Al

particle size

20-60 μm

pH

8.5-12.0

SMILES string

[Al].[Ni]

InChI

1S/Al.Ni

InChI key

NPXOKRUENSOPAO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Suitable as a hydrogenation catalyst for aromatic, olefin, nitrile and nitro group reductions.
Used as reducing agent for hydrogenation of organic compounds, e.g. hydrogenation of nitriles to amines, reduction of aromatic groups to the corresponding saturated compounds, hydrogenation reaction of β-keto esters, rreduction of amination, and also used as 3-alkylation of oxindole with alcohol, and Ullman-type cross coupling, and dehalogenation reactions.

Legal Information

Raney is a registered trademark of W.R. Grace & Co.-Conn.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Pyr. Liq. 1 - Skin Sens. 1 - STOT RE 1

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The shaping movement: fifth-generation technology.
Clifford J Ruddle et al.
Dentistry today, 32(4), 94-94 (2013-05-11)
Michał Sarul et al.
Advances in clinical and experimental medicine : official organ Wroclaw Medical University, 22(2), 253-260 (2013-05-28)
Cognizance of the mechanical properties of nickel-titanium archwires is necessary for the management of orthodontic therapy with fixed appliances. Acting on the periodontium with forces that are too heavy may lead to such complications as: pain, tooth root resorption and
Fariborz Amini et al.
International journal of orthodontics (Milwaukee, Wis.), 24(1), 43-44 (2013-06-05)
In this report, we present an uncomplicated approach to overcome lingual rolling of mandibular molars during orthodontic treatment, by using rectangular NiTi arch wire in twist fashion through the molar tube. The sequential steps of the twist manoeuvre on a
Palatally anchored maxillary molar mesialization using the mesialslider.
Benedict Wilmes et al.
Journal of clinical orthodontics : JCO, 47(3), 172-179 (2013-05-11)
A J Feilzer et al.
Nederlands tijdschrift voor tandheelkunde, 120(6), 335-341 (2013-07-19)
Orally applied metal alloys can cause undesirable physical effects. A distinction needs to be made in this respect between local and systemic reactions and toxic and immunological reactions. A case is presented which illustrates this problem. In this case, the

Articles

Nickel complexes catalyze various synthetic reactions like oxidative addition, C-H activation, and cross-coupling.

Nickel complexes catalyze various synthetic reactions like oxidative addition, C-H activation, and cross-coupling.

Nickel complexes catalyze various synthetic reactions like oxidative addition, C-H activation, and cross-coupling.

Nickel complexes catalyze various synthetic reactions like oxidative addition, C-H activation, and cross-coupling.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service