Skip to Content
Merck
All Photos(2)

Key Documents

45449

Supelco

Dimethoate

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H12NO3PS2
CAS Number:
Molecular Weight:
229.26
Beilstein:
1785339
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

CNC(CSP(OC)(OC)=S)=O

InChI

1S/C5H12NO3PS2/c1-6-5(7)4-12-10(11,8-2)9-3/h4H2,1-3H3,(H,6,7)

InChI key

MCWXGJITAZMZEV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Chronic 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

224.6 °F - closed cup

Flash Point(C)

107 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Nander Van Praet et al.
Aquatic toxicology (Amsterdam, Netherlands), 155, 236-243 (2014-07-27)
The combined effects of a pesticide and predation risk on sublethal endpoints in the midge Chironomus riparius were investigated using a combination of predator-release kairomones from common carp (Cyprinus carpio) and alarm substances from conspecifics together with the pesticide dimethoate.
Grażyna Wilczek et al.
Comparative biochemistry and physiology. Toxicology & pharmacology : CBP, 157(2), 119-131 (2012-10-27)
The aim of this study was to assess the intensity of enzymatic antioxidative parameters [i.e., superoxide dismutase (SOD), catalase (CAT), and the glutathione peroxidases each selene dependent, GPOX or selene independent, including GSTPx, glutathione S-transferase, and GST] and non-enzymatic antioxidative
Mirna Velki et al.
Comparative biochemistry and physiology. Toxicology & pharmacology : CBP, 156(2), 104-112 (2012-05-23)
Earthworms ingest large amounts of soil and therefore are continuously exposed to contaminants through their alimentary surfaces. Additionally, several studies have shown that earthworm skin is a significant route of contaminant uptake as well. In order to determine effects of
Mariana Astiz et al.
Toxicology and applied pharmacology, 272(2), 263-271 (2013-07-31)
Dimethoate is an organophosphorus insecticide extensively used in horticulture. Previous studies have shown that the administration of dimethoate to male rats, at a very low dose and during a sub-chronic period, increases the oxidation of lipids and proteins, reduces the
Suguru Yoshida et al.
Journal of the American Chemical Society, 134(47), 19358-19361 (2012-11-16)
A novel nucleophilic substitution reaction at the nitrogen of arylsulfonamides by means of phosphide anions has been described. This reaction allows for the efficient transformation of arylsulfonamides into synthetically valuable phosphamides, amines, and a variety of protected amines.

Protocols

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service