Skip to Content
Merck
All Photos(2)

Documents

45632

Supelco

Profenofos

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H15BrClO3PS
CAS Number:
Molecular Weight:
373.63
Beilstein:
2150258
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

CCCSP(=O)(OCC)Oc1ccc(Br)cc1Cl

InChI

1S/C11H15BrClO3PS/c1-3-7-18-17(14,15-4-2)16-11-6-5-9(12)8-10(11)13/h5-6,8H,3-4,7H2,1-2H3

InChI key

QYMMJNLHFKGANY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Profenofos is a broad-spectrum organophosphorus pesticide, commonly used for the control of various insect pests on vegetable crops.

Application

Profenofos may be used as an analytical reference standard for the quantification of the analyte in vegetables using high-performance liquid chromatography.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

255.2 °F - closed cup

Flash Point(C)

124 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Residual behaviour of profenofos on some field-grown vegetables and its removal using various washing solutions and household processing
Radwan.A.M, et al.
Food And Chemical Toxicology, 43(4), 553-557 (2005)
Determination of the organophosphorus pesticide in vegetables by high-performance liquid chromatography
Baig AS, et al.
American-Eurasian Journal of Agriculture and Environment Science, 6(5), 513-519 (2009)
Irani Mukherjee et al.
Bulletin of environmental contamination and toxicology, 88(3), 338-343 (2011-11-08)
Persistence behavior of two combination mix formulations of insecticides viz. profenofos + cypermethrin (Roket 44EC) and chlorpyrifos + cypermethrin (Nacraj-505) were studied in eggplant fruits following spray application at recommended and double dose. Half-life values were calculated from first order
P Kavitha et al.
Ecotoxicology and environmental safety, 72(6), 1727-1733 (2009-06-09)
The euryhaline fish, Oreochromis mossambicus was exposed to sub-lethal concentration (30 microg/L) of profenofos (PF) for 28 days and allowed to recover for 7 days. Responses were evaluated through antioxidant enzyme activities in various tissues of fish. Glutathione-S-transferase (GST) and
Kriengkrai Satapornvanit et al.
Chemosphere, 74(9), 1209-1215 (2008-12-24)
A bioassay was developed using post-larvae of freshwater prawn Macrobrachium rosenbergii (length 9-10 mm) in order to determine the toxicity of profenofos, dimethoate, chlorpyrifos, carbendazim and zinc. This was the first study in the tropics with M. rosenbergii, particularly at

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service