Skip to Content
Merck
All Photos(1)

Documents

36195

Supelco

Phosmet

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H12NO4PS2
CAS Number:
Molecular Weight:
317.32
Beilstein:
264869
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

COP(=S)(OC)SCN1C(=O)c2ccccc2C1=O

InChI

1S/C11H12NO4PS2/c1-15-17(18,16-2)19-7-12-10(13)8-5-3-4-6-9(8)11(12)14/h3-6H,7H2,1-2H3

InChI key

LMNZTLDVJIUSHT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Phosmet is a widely used, broad spectrum insecticide, which belongs to the phosphorodithioate subclass of organophosphates. It can find applications in having a control over insects in horticultural crops and ornamental plants. It can be used to control olive fruit fly (Bactrocera oleae, Dacus oleae), which can cause damage to the olive oil trees.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 2 - STOT SE 1

Target Organs

Nervous system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Jaya Sitjar et al.
Nanomaterials (Basel, Switzerland), 9(5) (2019-05-01)
Health risks posed by the exposure to trace amounts of pesticide residue in agricultural products have gained a lot of concerns, due to their neurotoxic nature. The applications of surface-enhanced Raman Scattering (SERS) as a detection technique have consistently shown
Analyses of phosmet residues in apples with surface-enhanced Raman spectroscopy
Fan Y, et al.
Food Control, 37, 153-157 (2014)
osmet and its metabolites in olives by matrix solid-phase dispersion and gas chromatography?mass spectrometry
Cunha.CS, et al.
Talanta, 73, 514-522 (2007)
Jaya Sitjar et al.
Nanomaterials (Basel, Switzerland), 9(12) (2019-12-15)
Nanostructures with spikes (NSPs) have been a subject of several surface-enhanced Raman scattering (SERS) applications owing to their significant Raman signal enhancement brought about by the combined effects of interspike coupling and the accumulated induction on the tips of spikes.
Katrin Sinderhauf et al.
Journal of agricultural and food chemistry, 52(26), 8046-8052 (2004-12-23)
The organophosphorus insecticide phosmet is commonly used for plant protection as well as against pests on animals. Phosmet features numerous degradation pathways induced by UV irradiation. In this study, we focused on the reaction possibilities of phosmet in the presence

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service