440108
Trichloro(phenyl)silane
≥97.0%
Synonym(s):
Phenyltrichlorosilane
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Quality Level
Assay
≥97.0%
form
liquid
refractive index
n20/D 1.523 (lit.)
bp
201 °C (lit.)
density
1.321 g/mL at 25 °C (lit.)
SMILES string
Cl[Si](Cl)(Cl)c1ccccc1
InChI
1S/C6H5Cl3Si/c7-10(8,9)6-4-2-1-3-5-6/h1-5H
InChI key
ORVMIVQULIKXCP-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Trifluoro(phenyl)silane can react with aliphatic alcohols to yield benzene and alkoxyfluoro and tetraalkoxysilanes. Hydrolytic condensation of this silane in its vapor phase results in the formation of 1,1,3,3-Tetrachloro-1,3-diphenyldisiloxane.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Eye Dam. 1 - Skin Corr. 1A
Supplementary Hazards
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 1
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Cleavage of the C-Si bond in Trifluoro (phenyl) silane with Aliphatic Alcohols.
Russ. J. Gen. Chem., 75(12), 1927-1929 (2005)
Journal of hazardous materials, 336, 222-231 (2017-05-12)
In this study, a novel strategy was developed to fabricate highly flame retardant polymer foam composite materials coated by synthesized silicone resin (SiR) polymer via a facile dip-coating processing. Applying the SiR polymer coating, the mechanical property and thermal stability
Syntheses and structural analyses of 1, 3-diphenyldisiloxanes as a building block of silsesquioxanes.
Bulletin of the Chemical Society of Japan, 76(10), 1983-19887 (2003)
Journal of colloid and interface science, 547, 145-152 (2019-04-06)
Wicking and percolation of a liquid in porous media is, among other factors, strongly affected by wettability of the pores. Nitrocellulose (NC) fibrous matrix impregnated with nitroglycerin (NG) is a main component of propellant formulations. Over time, NG, being a
Beilstein journal of nanotechnology, 9, 1211-1219 (2018-05-17)
Nanostructures of 4-(chloromethyl)phenyltrichlorosilane (CMPS) were used as a foundation to attach and grow heterostructures of porphyrins and organosilanes. A protocol was developed with particle lithography using steps of immersion in organosilane solutions to selectively passivate the surface of Si(111) with
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service