Skip to Content
Merck
All Photos(3)

Key Documents

104876

Sigma-Aldrich

Trichlorovinylsilane

97%

Synonym(s):

(Trichlorosilyl)ethylene, Vinyltrichlorosilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHSiCl3
CAS Number:
Molecular Weight:
161.49
Beilstein:
1743440
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

vapor density

>1 (vs air)

vapor pressure

60 mmHg ( 23 °C)

Assay

97%

refractive index

n20/D 1.436 (lit.)

bp

90 °C (lit.)

mp

−95 °C (lit.)

density

1.27 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

Cl[Si](Cl)(Cl)C=C

InChI

1S/C2H3Cl3Si/c1-2-6(3,4)5/h2H,1H2

InChI key

GQIUQDDJKHLHTB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Trichlorovinylsilane may react with lithium-tert-butyl, and 1,3-enynes to yield isomeric silacyclobutanes and silacyclobutenes respectively.

Application

A study reports the possible use of this silane for treating Pluronic F127, chitosan for better attachment and proliferation of endothelial cells on biomaterials. Trichlorovinylsilane may be used to prepare epoxy terminated carbosiloxane.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

50.0 °F - closed cup

Flash Point(C)

10 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Micropatterning of proteins and mammalian cells on biomaterials.
Faseb Journal, 18(3), 525-527 (2004)
Xiangming Wu et al.
Colloids and surfaces. B, Biointerfaces, 130, 69-76 (2015-04-25)
Earlier work provided direction for development of responsive drug delivery systems based on modulation of the structure, amphiphilicity, and surface density of bioactive peptides entrapped within pendant polyethylene oxide (PEO) brush layers. In this work, we describe the sequential and
Synthesis, characterization of an epoxy?terminated carbosiloxane and its application as a modifier in epoxy systems.
Polym. Eng. Sci., 52(4), 868-874 (2012)
John D DeSain et al.
Physical chemistry chemical physics : PCCP, 8(19), 2240-2248 (2006-05-12)
The absolute gas phase ultraviolet absorption spectra of trichlorovinylsilane and allyltrichlorosilane have been measured from 191 to 220 nm. Over this region the absorption spectra of both species are broad and relatively featureless, and their cross sections increase with decreasing
Trichlorovinylsilane, lithium-tert-butyl, and 1, 3-enynes: A versatile combination for the competitive formation of silacyclobutanes and silacyclobutenes.
Canadian Journal of Chemistry, 78(11), 1445-1458 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service