104744
Trimethoxyphenylsilane
97%
Synonym(s):
(Trimethoxysilyl)benzene, Phenyltrimethoxysilane
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
C6H5Si(OCH3)3
CAS Number:
Molecular Weight:
198.29
Beilstein:
2937896
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
Quality Level
Assay
97%
form
liquid
refractive index
n20/D 1.468 (lit.)
bp
233 °C (lit.)
density
1.062 g/mL at 25 °C (lit.)
SMILES string
CO[Si](OC)(OC)c1ccccc1
InChI
1S/C9H14O3Si/c1-10-13(11-2,12-3)9-7-5-4-6-8-9/h4-8H,1-3H3
InChI key
ZNOCGWVLWPVKAO-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Trimethoxyphenylsilane (TMPS) is an organotrialkoxysilane that is used as a functional monomer in the sol-gel processing.
Application
TMPS can also be used in the preparation of polytrimethoxyphenylsilane (PTMS) that facilitates the removal of graphene after chemical vapor deposition (CVD). Molecularly imprinted sol gels can be synthesized by using TMPS for the fabrication of localized surface plasmon resonance (LSPR).
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Flam. Liq. 3 - STOT RE 2 Oral
Target Organs
Bladder
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
84.2 °F - closed cup
Flash Point(C)
29 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Bio-inspired molecularly imprinted polymer-green emitting carbon dot composite for selective and sensitive detection of 3-nitrotyrosine as a biomarker
Jalili R and Amjadi M
Sensors and Actuators B, Chemical, 255, 1072-1078 (2018)
Ti-POSS covalently immobilized onto mesoporous silica: A model for active sites in heterogeneous catalytic epoxidation
Carniato, F., Bisio, C., Sordelli, L., Gavrilova, E., & Guidotti, M.
Inorgorganica Chimica Acta, 380, 244-251 (2012)
Capillary microextraction of volatiles device for enhanced BTEX vapors sampling based on a phenyl modified PDMS sol-gel adsorption phase
Gura S, et al.
Analytica Chimica Acta, 1014, 27-40 (2018)
Xiaomei Yang et al.
Materials (Basel, Switzerland), 12(12) (2019-06-20)
In this work, we reported the preparation and chemical analysis of novel polyhedral oligomeric silsesquioxane (POSS)-based porous materials, which were prepared according to Friedel-Crafts chloromethylation by using aluminum chloride as the catalyst and dichloromethane as the solvent. Through controlling the
Poly-Trimethoxyphenylsilane as Carrier Film for Residual-Free CVD Graphene Transfer
Kim BJ, et al.
Physica Status Solidi - Rapid Research Letters, 11(11), 1700240-1700240 (2017)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service