Skip to Content
Merck
All Photos(4)

Key Documents

B56307

Sigma-Aldrich

Bromoacetic acid

reagent grade, 97%

Synonym(s):

2-Bromoethanoic acid, Bromoethanoic acid, Carboxymethyl bromide, MBAA, Monobromoacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrCH2COOH
CAS Number:
Molecular Weight:
138.95
Beilstein:
506167
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

grade

reagent grade

Assay

97%

form

crystals

bp

208 °C (lit.)

mp

47-49 °C (lit.)

SMILES string

OC(=O)CBr

InChI

1S/C2H3BrO2/c3-1-2(4)5/h1H2,(H,4,5)

InChI key

KDPAWGWELVVRCH-UHFFFAOYSA-N

Gene Information

human ... PTPN6(5777)

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Bromoacetic acid is commonly used in acylation, alkylation, condensation, addition, cycloaddition, and esterification reactions.

Application

Bromoacetic acid can be used:
  • As a quaternizing and alkylating agent.
  • For carboxymethylation of poly(ethylene glycol) for bacterial adhesion experiments.
  • For the synthesis of C60-fused lactone in the presence of Mn(OAc)3·2H2O.

related product

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Oral - Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Aquatic Acute 1 - Eye Dam. 1 - Skin Corr. 1A - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bromoacetic Acid
Nataliya K and Mikhailo S
eEROS (Encyclopedia of Reagents for Organic Synthesis) (2022)
Synthesis of new cationics, carboxybetaines, and sulfobetaines Bolaamphiphiles.
Souirti S and Baboule M
Synthetic Communications, 31(1), 9-18 (2001)
Covalent attachment of poly (ethylene glycol) to surfaces, critical for reducing bacterial adhesion.
Kingshott P, et al.
Langmuir, 19(17), 6912-6921 (2003)
Manganese (III) acetate-mediated radical reaction of [60] fullerene with bromoacetic acid, 3-chloropropionic acid or 1-naphthylacetic acid.
Li F, et al.
Chinese Science Bulletin, 55(25), 2909-2914 (2010)
A selective fluorescent sensor for detecting lead in living cells.
He Q, et al.
Journal of the American Chemical Society, 128(29), 9316-9317 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service