Skip to Content
Merck
All Photos(1)

Key Documents

38990

Sigma-Aldrich

2-Dimethylaminoethanol

purum, ≥98.0% (GC)

Synonym(s):

Deanol, N,N-Dimethyl-2-hydroxyethylamine, N,N-Dimethylethanolamine, Jeffcat DMEA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NCH2CH2OH
CAS Number:
Molecular Weight:
89.14
Beilstein:
1209235
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

100 mmHg ( 55 °C)
6.12 mmHg ( 20 °C)

Quality Level

grade

purum

Assay

≥98.0% (GC)

refractive index

n20/D 1.4294 (lit.)
n20/D 1.430

bp

134-136 °C (lit.)

mp

−70 °C (lit.)

solubility

H2O: soluble

density

0.886 g/mL at 20 °C (lit.)

SMILES string

CN(C)CCO

InChI

1S/C4H11NO/c1-5(2)3-4-6/h6H,3-4H2,1-2H3

InChI key

UEEJHVSXFDXPFK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

2-Dimethylaminoethanol (Deanol) can be used to functionalize the sensitizers in dye-sensitized solar cells to prevent aggregation and to increase the dye adsorption density. It is used in the synthesis of NNO-tridentate Schiff base ligands to prepare zinc alkoxide complexes that catalyze ring-opening polymerization of L-lactide. It is also used in the preparation of ionic liquids.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

104.0 °F - Seta closed cup

Flash Point(C)

40 °C - Seta closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Molecular engineering of zinc phthalocyanine sensitizers for efficient dye-sensitized solar cells.
Ikeuchi, Takuro et al.
Chemical Communications (Cambridge, England), 50(16), 1941-1943 (2014)
Dual functions of N, N-dimethylethanolamnium-based ionic liquids for the Knoevenagel reactions at room temperature.
Zhu, Anlian et al.
Catalysis Today, 200(11), 17-23 (2013)
Ring-opening polymerization of lactides initiated by zinc alkoxides derived from NNO-tridentate ligands.
Chen H Y, et al.
Macromolecules, 39(11), 3745-3752 (2006)
B Sommerfeld
Phytomedicine : international journal of phytotherapy and phytopharmacology, 14(11), 711-715 (2007-10-10)
Tricutan is a combination product of herbal extracts traditionally used for treatment of skin conditions, together with dimethylaminoethanol. The effectiveness of Tricutan in improving skin firmness and elasticity in photoaged facial skin was examined in a randomised, placebo-controlled, double-blind, split-face
Tadashi Tatsumi et al.
Bioorganic & medicinal chemistry, 18(7), 2720-2727 (2010-03-17)
Effects of retro-inverso (RI) modifications of HTLV-1 protease inhibitors containing a hydroxyethylamine isoster backbone were clarified. Construction of the isoster backbone was achieved by a stereoselective aldol reaction. Four diastereomers with different configurations at the isoster hydroxyl site and the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service