Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
2-Dimethylaminoethanol
Synonym(s):
N
,
N
-Dimethyl-2-hydroxyethylamine,
N
,
N
-Dimethylethanolamine, Jeffcat DMEA, 2-(Dimethylamino)-ethanol
Linear Formula:
(CH
3
)
2
NCH
2
CH
2
OH
CAS Number:
108-01-0
Molecular Weight:
89.14
EC Number:
203-542-8
Products
Product Category
building blocks (3)
Brand
Sigma-Aldrich (4)
Supelco (1)
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Purum (1)
analytical standard (1)
Technique
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
471453
2-Dimethylaminoethanol,
≥99.5%
Expand
View Pricing
8.03237
2-Dimethylaminoethanol,
for synthesis
Expand
View Pricing
391263
2-Dimethylaminoethanol,
purified by redistillation, ≥99.5%
Expand
View Pricing
38990
2-Dimethylaminoethanol,
purum, ≥98.0% (GC)
Expand
View Pricing
50632
2-Dimethylaminoethanol,
analytical standard
Expand
View Pricing