Skip to Content
Merck
All Photos(1)

Key Documents

471445

Sigma-Aldrich

2-(Methylamino)ethanol

≥98%

Synonym(s):

N-Methylethanolamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3NHCH2CH2OH
CAS Number:
Molecular Weight:
75.11
Beilstein:
1071196
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

2.6 (vs air)

vapor pressure

0.5 mmHg ( 20 °C)

Assay

≥98%

form

liquid

autoignition temp.

662 °F

expl. lim.

19.8 %

refractive index

n20/D 1.439 (lit.)

bp

159 °C (lit.)

density

0.935 g/mL at 25 °C (lit.)

functional group

amine
hydroxyl

SMILES string

CNCCO

InChI

1S/C3H9NO/c1-4-2-3-5/h4-5H,2-3H2,1H3

InChI key

OPKOKAMJFNKNAS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

2-(Methylamino)ethanol is used as an organic building block in chemical synthesis. It also finds applications in various fields such as textile lubricants, polishes, detergents, and in personal care products. In addition to this, it is used as a solvent for the removal of carbon dioxide from the gas stream.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Repr. 2 - Skin Corr. 1B - STOT RE 2 Oral - STOT SE 3

Target Organs

Kidney,Testes,Liver,spleen,ovary, Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

168.8 °F - closed cup

Flash Point(C)

76 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrzej Grajkowski et al.
Nucleic acids research, 33(11), 3550-3560 (2005-06-24)
A CpG-containing DNA oligonucleotide functionalized with the 2-(N-formyl-N-methyl)aminoethyl thiophosphate protecting group (CpG ODN fma1555) was prepared from phosphoramidites 1a-d using solid-phase techniques. The oligonucleotide behaved as a prodrug by virtue of its conversion to the well-studied immunomodulatory CpG ODN 1555
Volumetric properties, viscosities, and refractive indices for aqueous 2-(Methylamino) ethanol solutions from (298.15 to 343.15) K.
Li J, et al.
Journal of Chemical and Engineering Data, 42(2), 560-565 (2007)
A ?core-shell? approach to producing 3D polymer nanocomposites.
Kalinina O and Kumacheva E.
Macromolecules, 32(12), 4122-4129 (1999)
Roberto Motterlini et al.
Redox biology, 20, 334-348 (2018-11-06)
Oxidative stress and inflammation are predominant features of several chronic diseases. The nuclear factor erythroid 2-related factor 2 (Nrf2) is a major arbiter in counteracting these insults via up-regulation of several defensive proteins, including heme oxygenase-1 (HO-1). HO-1-derived carbon monoxide
Densities and Viscosities of Aqueous Ternary Mixtures of 2-(Methylamino) ethanol and 2-(Ethylamino) ethanol with Diethanolamine, Triethanolamine, N-Methyldiethanolamine, or 2-Amino-1-methyl-1-propanol from 298.15 to 323.15 K.
Alvarez e, et al.
Journal of Chemical and Engineering Data, 51(3), 955-962 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service