36631
Atrazine-2-hydroxy
PESTANAL®, analytical standard
Synonym(s):
2-Hydroxyatrazine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C8H15N5O
CAS Number:
Molecular Weight:
197.24
Beilstein:
5870987
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Recommended Products
grade
analytical standard
Quality Level
product line
PESTANAL®
shelf life
limited shelf life, expiry date on the label
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
agriculture
environmental
format
neat
SMILES string
CCNc1nc(O)nc(NC(C)C)n1
InChI
1S/C8H15N5O/c1-4-9-6-11-7(10-5(2)3)13-8(14)12-6/h5H,4H2,1-3H3,(H3,9,10,11,12,13,14)
InChI key
NFMIMWNQWAWNDW-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Atrazine-2-hydroxy is a non-herbicidal, primary photoproduct obtained upon hydrolytic dechlorination of atrazine.
Application
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Legal Information
PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Anaerobic degradation of atrazine
Boopathy R, et al.
International Biodeterioration & Biodegradation, 119, 626-630 (2017)
The Triazine Herbicides (2011)
Milan Hutta et al.
Journal of separation science, 29(13), 1977-1987 (2006-10-05)
A well established method of direct injection of larger than conventional sample volumes ranging from 0.1 mL to 10 mL in HPLC is the injection valve method in which a loop of tubing is totally or partially filled with sample.
J Mahía et al.
Journal of environmental quality, 36(3), 826-831 (2007-05-09)
Mineralization of atrazine and formation of extractable and non-extractable "bound" residues were followed under laboratory conditions in two contrasting soils (organic C, texture, and atrazine application history) from northern Spain. The soils, a Humic Cambisol (MP) and a Gleyic Cambisol
Susan C Laws et al.
Toxicological sciences : an official journal of the Society of Toxicology, 76(1), 190-200 (2003-09-13)
We showed previously that the chlorotriazine herbicide, atrazine (ATR), delays the onset of pubertal development in female rats. ATR and its biotransformation by-products are present in soil and groundwater. Since current maximum contaminant levels are set only for ATR, it
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service