Skip to Content
Merck
All Photos(1)

Key Documents

36628

Supelco

Atrazine-desisopropyl

PESTANAL®, analytical standard

Synonym(s):

2-Amino-4-chloro-6-ethylamino-1,3,5-triazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H8ClN5
CAS Number:
Molecular Weight:
173.60
Beilstein:
8150
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable
solid phase extraction (SPE): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCNc1nc(N)nc(Cl)n1

InChI

1S/C5H8ClN5/c1-2-8-5-10-3(6)9-4(7)11-5/h2H2,1H3,(H3,7,8,9,10,11)

InChI key

IVENSCMCQBJAKW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Atrazine-desisopropyl is commonly used herbicide, which belongs to the class of triazines and is found to be a metabolite of atrazine.
Standard for Supelco MIP SPE cartridges. For more information request Supelco Literature T407075, T706023

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Determination of atrazine, desethyl atrazine and desisopropyl atrazine in environmental water samples using hollow fiber-protected liquid-phase microextraction and high performance liquid chromatography
Peng J, et al.
Microchimica Acta, 158(1), 181-186 (2007)
D Q Tran et al.
Biochemical and biophysical research communications, 227(1), 140-146 (1996-10-03)
The chloro-S-triazine derived compounds atrazine, atrazine desisopropyl, cyanazine, and simazine are commonly used herbicides. These compounds do not have estrogenic activity in yeast expressing human estrogen receptor (hER) and an estrogen-sensitive reporter. In the presence of a concentration of estradiol
R Ikonen et al.
Toxicology letters, 44(1-2), 109-112 (1988-11-01)
Rats were given atrazine (2-chloro-4-ethylamino-6-(isopropylamino)-s-triazine) in drinking water for 1 or 3 weeks at 0.1 (0.45 mM), 0.2 (0.9 mM) or 0.5 g/l (2.3 mM) concentrations of the commercial agent. They excreted at both time points as the principal metabolite
K Wenger et al.
Journal of environmental quality, 34(6), 2187-2196 (2005-11-09)
DIMBOA (3,4-dihydro-2,4-dihydroxy-7-methoxy-2H-1,4-benzoxazin-3-one), a major benzoxazinone of Poaceae plants, was isolated and purified from corn seedlings. The effect of isolated and purified DIMBOA on the degradation of atrazine [2-chloro-4-(ethylamino)-6-(isopropylamino)-s-triazine], and its toxic breakdown products, desethylatrazine [2-chloro-4-amino-6-(isopropylamino)-s-triazine; DEA] and desisopropylatrazine [2-chloro-4-(ethylamino)-6-amino-s-triazine; DIA]
Z Q Shao et al.
Journal of bacteriology, 177(20), 5748-5755 (1995-10-01)
We used degenerate oligodeoxyribonucleotides derived from the N-terminal sequence of the s-triazine hydrolase from Rhodococcus corallinus NRRL B-15444R in an amplification reaction to isolate a DNA segment containing a 57-bp fragment from the trzA gene. By using the nucleotide sequence

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service