Skip to Content
Merck
All Photos(1)

Key Documents

C33301

Sigma-Aldrich

2-Chloro-4,6-diamino-1,3,5-triazine

95%

Synonym(s):

6-Chloro-2,4-diamino-1,3,5-triazine, Desethyl-desisopropyl-atrazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H4ClN5
CAS Number:
Molecular Weight:
145.55
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

95%

form

powder

mp

>320 °C (lit.)

SMILES string

Nc1nc(N)nc(Cl)n1

InChI

1S/C3H4ClN5/c4-1-7-2(5)9-3(6)8-1/h(H4,5,6,7,8,9)

InChI key

FVFVNNKYKYZTJU-UHFFFAOYSA-N

Gene Information

mouse ... Esr1(13982)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Greg P Dooley et al.
Chemical research in toxicology, 19(5), 692-700 (2006-05-16)
Atrazine (2-chloro-4-[ethylamino]-6-[isopropylamino]-1,3,5-triazine) is one of the most commonly used herbicides in North America and is frequently detected in ground and surface waters. This research investigated possible covalent modifications of hemoglobin following in vivo exposures to atrazine in Sprague Dawley (SD)
Jennifer L Seffernick et al.
Applied and environmental microbiology, 68(9), 4672-4675 (2002-08-30)
2-Chloro-4,6-diamino-s-triazine (CAAT) is a metabolite of atrazine biodegradation in soils. Atrazine chlorohydrolase (AtzA) catalyzes the dechlorination of atrazine but is unreactive with CAAT. In this study, melamine deaminase (TriA), which is 98% identical to AtzA, catalyzed deamination of CAAT to
Joseane M Pozzebon et al.
Journal of chromatography. A, 987(1-2), 375-380 (2003-03-05)
A method was developed to determine simazine, atrazine and their metabolite, 2-chloro-4,6-diamino-1,3,5-triazine, in urine. The presence of these herbicides in urine may reflect possible exposure to pesticides. Sample preparation involved protein precipitation and solid-phase extraction. The samples were analyzed by
C Ernst et al.
Applied microbiology and biotechnology, 42(5), 763-768 (1995-01-01)
A bacterium utilizing 2-chloro-4,6-diamino-s-triazine (CAAT) as sole nitrogen source was isolated under a N2-free atmosphere and identified as Klebsiella pneumoniae. Concomitant to CAAT degradation the protein content increased and chloride was released into the medium. Under air and a N2-atmosphere
Susan C Laws et al.
Toxicological sciences : an official journal of the Society of Toxicology, 76(1), 190-200 (2003-09-13)
We showed previously that the chlorotriazine herbicide, atrazine (ATR), delays the onset of pubertal development in female rats. ATR and its biotransformation by-products are present in soil and groundwater. Since current maximum contaminant levels are set only for ATR, it

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service