Skip to Content
Merck
All Photos(1)

Key Documents

34389

Supelco

Cyprodinil

PESTANAL®, analytical standard

Synonym(s):

4-Cyclopropyl-6-methyl-N-phenylpyrimidin-2-amine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H15N3
CAS Number:
Molecular Weight:
225.29
Beilstein:
7813601
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

Cc1cc(nc(Nc2ccccc2)n1)C3CC3

InChI

1S/C14H15N3/c1-10-9-13(11-7-8-11)17-14(15-10)16-12-5-3-2-4-6-12/h2-6,9,11H,7-8H2,1H3,(H,15,16,17)

InChI key

HAORKNGNJCEJBX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Coraline Mattei et al.
Chemosphere, 228, 75-82 (2019-04-26)
A fraction of the atmospheric pesticides can be adsorbed on particles surface according to their physicochemical properties. After adsorption, pesticides can undergo heterogeneous reactivity with atmospheric oxidants such as ozone, but the influence of the pesticide surface coating (i.e., the
H Basa Cesnik et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 25(4), 438-443 (2008-03-19)
Although the list of pesticides used in integrated pest management (IPM) in grape growing and their annual application rates are limited, we are still confronted with the problem of pesticide residues in grapes. This paper presents the results of pesticide
G A Bardas et al.
Phytopathology, 98(4), 443-450 (2008-10-24)
The fitness of anilinopyrimidine-resistant isolates of Botrytis cinerea compared with that of sensitive isolates, collected from vegetable crops in Greece during 2005, was investigated. Stability of resistance to anilinopyrimidine fungicides was determined after consecutive transfers of the fungal isolates on
Arnaud Auber et al.
Ecotoxicology (London, England), 20(8), 2042-2055 (2011-07-28)
The impacts of current and alternative wheat crop protection programs were compared in outdoor pond mesocosms in a 10-month long study. Realistic exposure scenarios were built based upon the results of modelling of drift, drainage and runoff of pesticides successively
Gustavo Cordero-Bueso et al.
Frontiers in microbiology, 8, 2025-2025 (2017-11-23)
The increasing level of hazardous residues in the environment and food chains has led the European Union to restrict the use of chemical fungicides. Thus, exploiting new natural antagonistic microorganisms against fungal diseases could serve the agricultural production to reduce

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service