Skip to Content
Merck
All Photos(1)

Key Documents

36531

Supelco

Difenoconazol

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H17Cl2N3O3
CAS Number:
Molecular Weight:
406.26
Beilstein:
9073356
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CC1COC(Cn2cncn2)(O1)c3ccc(Oc4ccc(Cl)cc4)cc3Cl

InChI

1S/C19H17Cl2N3O3/c1-13-9-25-19(27-13,10-24-12-22-11-23-24)17-7-6-16(8-18(17)21)26-15-4-2-14(20)3-5-15/h2-8,11-13H,9-10H2,1H3

InChI key

BQYJATMQXGBDHF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Difenoconazol is a broad spectrum, triazole fungicide, which can serve as a potent inhibitor of styrol.(1) It can be widely used in the protection of grain crops, fruits and sugar beet.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Wang et al.
Ecotoxicology and environmental safety, 86, 111-115 (2012-10-16)
An analytical method for determining the residue of a broad-spectrum fungicide, difenoconazole, in soil and rice crop matrices is described. Mean recoveries and relative standard deviation (RSD) in paddy soil, water, rice plant, rice hull, and husked rice matrices at
Kaushik Banerjee et al.
Pest management science, 64(3), 283-289 (2008-01-18)
Tetraconazole and difenoconazole are triazole fungicides with proven bioefficacy against grapevine powdery mildew disease. In the present work, the authors explored the residue dynamics of these two compounds in grapes and determined their preharvest intervals (PHIs) corresponding to multiple field
Occurrence and origin of sensitivity toward difenoconazole in zebrafish (Danio reio) during different life stages
Mu X, et al.
Aquatic Toxicology (Amsterdam, Netherlands), 57-68 (2015)
Tom R Bilbo et al.
Journal of economic entomology, 113(5), 2096-2103 (2020-08-03)
The twospotted spider mite (TSSM, Tetranychus urticae Koch) is a key pest of tomatoes (Solanum lycopersicon L. [Solanales: Solanaceae]) in North Carolina, and its management has relied principally on synthetic acaricides. Augmentative biological control of TSSM is a commonplace and
Livia Pitombeira de Figueirêdo et al.
Environmental pollution (Barking, Essex : 1987), 244, 342-350 (2018-10-24)
Pesticides risk assessments have traditionally focused on the effects on standard parameters, such as mortality, reproduction and development. However, one of the first signs of adverse effects that occur in organisms exposed to stress conditions is an alteration in their

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service