Skip to Content
Merck
All Photos(1)

Documents

33696

Supelco

Pyraclostrobin

PESTANAL®, analytical standard

Synonym(s):

Methyl {2-[1-(4-chlorophenyl)-1H-pyrazol-3-yloxymethyl]phenyl}methoxycarbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H18ClN3O4
CAS Number:
Molecular Weight:
387.82
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CON(C(=O)OC)c1ccccc1COc2ccn(n2)-c3ccc(Cl)cc3

InChI

1S/C19H18ClN3O4/c1-25-19(24)23(26-2)17-6-4-3-5-14(17)13-27-18-11-12-22(21-18)16-9-7-15(20)8-10-16/h3-12H,13H2,1-2H3

InChI key

HZRSNVGNWUDEFX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pyraclostrobin can be called as a broad spectrum fungicide, used to preserve high valuable crops, having preventive and curative properties and a long residual action.
Pyraclostrobin can be called as a broad spectrum fungicide, used to preserve high valuable crops having preventive effect, curative properties and a long residual action.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Handbook of Pesticide Toxicology, Two-Volume Set: Principles and Agents, Volume 1 (2001)
Krieger R et al.
Handbook of Pesticide Toxicology, Two-Volume Set: Principles and Agents, Volume 1, 1(2) (2001)
Production and characterization of monoclonal antibodies specific to the strobilurin pesticide pyraclostrobin
Mercader.VJ, et al.
Journal of Agricultural and Food Chemistry, 56, 7682-7690 (2008)
Development of immunoaffinity columns for pyraclostrobin extraction from fruit juices and analysis by liquid chromatography with UV detection
Esteve-Turrillas. AF, et al
Journal of Chromatography A, 1218, 4902-4909 (2011)
Development of immunoaffinity columns for pyraclostrobin extraction from fruit juices and analysis by liquid chromatography with UV detection
Esteve-Turrillas. AF, et al et al.
Journal of Chromatography A, 1218(30), 4902-4909 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service