Skip to Content
Merck
All Photos(3)

Documents

328111

Sigma-Aldrich

Phenol

unstabilized, purified by redistillation, ≥99%

Synonym(s):

Hydroxybenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5OH
CAS Number:
Molecular Weight:
94.11
Beilstein:
969616
EC Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
NACRES:
NA.21

vapor density

3.24 (vs air)

Quality Level

vapor pressure

0.09 psi ( 55 °C)
0.36 mmHg ( 20 °C)

Assay

≥99%

form

crystalline (solid)
crystals
liquid (after melt)

autoignition temp.

1319 °F

purified by

redistillation

expl. lim.

8.6 %

bp

181 °C
182 °C (lit.)

mp

40-42 °C (lit.)

solubility

water: soluble 84 g/L at 20 °C

density

1.071 g/mL at 25 °C (lit.)

SMILES string

Oc1ccccc1

InChI

1S/C6H6O/c7-6-4-2-1-3-5-6/h1-5,7H

InChI key

ISWSIDIOOBJBQZ-UHFFFAOYSA-N

Gene Information

human ... GABRA1(2554)

Looking for similar products? Visit Product Comparison Guide

General description

Phenol is the simplest aromatic alcohol building block.

Application

Phenol may be used as a phase-transfer catalyst in electrolyte solutions of the lithium-O2 battery to promote solution phase discharge.

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Muta. 2 - Skin Corr. 1B - STOT RE 2

Target Organs

Nervous system,Kidney,Liver,Skin

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

174.2 °F - closed cup

Flash Point(C)

79.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phenol?Catalyzed Discharge in the Aprotic Lithium?Oxygen Battery.
Gao X, et al.
Angewandte Chemie (Weinheim an der Bergstrasse, Germany) (2017)
Tyman JHP.
Synthetic and Natural Phenols., 67-67 (1996)
Eagleson M.
Concise Encyclopedia Chemistry, 812-812 (1994)
PREPARATION OF TRANSFORMING DEOXYRIBONUCLEIC ACID BY PHENOL TREATMENT.
H SAITO et al.
Biochimica et biophysica acta, 72, 619-629 (1963-08-20)
Tyman JHP.
Synthetic and Natural Phenols., 1-1 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service