Skip to Content
Merck
All Photos(1)

Key Documents

W322318

Sigma-Aldrich

Phenol

≥99%

Synonym(s):

Hydroxybenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5OH
CAS Number:
Molecular Weight:
94.11
FEMA Number:
3223
Beilstein:
969616
EC Number:
Council of Europe no.:
11811
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
4.041
NACRES:
NA.21
grade:
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens
Pricing and availability is not currently available.

biological source

synthetic

Quality Level

grade

Halal
Kosher

Agency

meets purity specifications of JECFA

vapor density

3.24 (vs air)

vapor pressure

0.09 psi ( 55 °C)
0.36 mmHg ( 20 °C)

Assay

≥99%

form

crystalline powder

autoignition temp.

1319 °F

may contain

≤0.15% hypophosphorous acid as additive

expl. lim.

8.6 %

bp

182 °C (lit.)

mp

40-42 °C (lit.)

density

1.071 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

phenolic

SMILES string

Oc1ccccc1

InChI

1S/C6H6O/c7-6-4-2-1-3-5-6/h1-5,7H

InChI key

ISWSIDIOOBJBQZ-UHFFFAOYSA-N

Gene Information

human ... GABRA1(2554)

Looking for similar products? Visit Product Comparison Guide

Application


  • Revealing mechanism of phenolic-amine reaction to form humus in compost based on high-resolution liquid chromatography mass spectrometry and spectroscopy.: This study delves into the complex reactions between phenolic compounds and amines during compost formation, shedding light on the molecular pathways that lead to humus production, crucial for soil fertility and carbon sequestration (Zhou et al., 2024).

  • Unlocking the potential of phenolated kraft lignin as a versatile feed additive.: This paper explores the utilization of phenolated kraft lignin as a feed additive, examining its benefits in animal nutrition and its role in sustainable agricultural practices (Li et al., 2024).

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Muta. 2 - Skin Corr. 1B - STOT RE 2

Target Organs

Nervous system,Kidney,Liver,Skin

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

177.8 °F - closed cup

Flash Point(C)

81 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mark J Lee et al.
PLoS pathogens, 11(10), e1005187-e1005187 (2015-10-23)
Of the over 250 Aspergillus species, Aspergillus fumigatus accounts for up to 80% of invasive human infections. A. fumigatus produces galactosaminogalactan (GAG), an exopolysaccharide composed of galactose and N-acetyl-galactosamine (GalNAc) that mediates adherence and is required for full virulence. Less
Thitiporn Anunthawan et al.
Biochimica et biophysica acta, 1848(6), 1352-1358 (2015-03-15)
We investigated the mechanisms of two tryptophan-rich antibacterial peptides (KT2 and RT2) obtained in a previous optimization screen for increased killing of both Gram-negative and Gram-positive bacteria pathogens. At their minimal inhibitory concentrations (MICs), these peptides completely killed cells of
Rita A Busuttil et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 20(10), 2761-2772 (2014-03-25)
Gene-expression profiling has revolutionized the way we think about cancer and confers the ability to observe the synchronous expression of thousands of genes. The use of putative genome-level expression profiles has allowed biologists to observe the complex interactions of genes
Saori Tomita et al.
Nature communications, 6, 6966-6966 (2015-04-30)
Estrogen receptor-α (ER)-positive breast cancer cells undergo hormone-independent proliferation after deprivation of oestrogen, leading to endocrine therapy resistance. Up-regulation of the ER gene (ESR1) is critical for this process, but the underlying mechanisms remain unclear. Here we show that the
Souhaila Bouatra et al.
PloS one, 8(9), e73076-e73076 (2013-09-12)
Urine has long been a "favored" biofluid among metabolomics researchers. It is sterile, easy-to-obtain in large volumes, largely free from interfering proteins or lipids and chemically complex. However, this chemical complexity has also made urine a particularly difficult substrate to

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service