Skip to Content
Merck
All Photos(1)

Key Documents

32606

Supelco

Fumonisin B3 solution

~50 μg/mL in acetonitrile: water, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H59NO14
CAS Number:
Molecular Weight:
705.83
Beilstein:
8181181
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

concentration

~50 μg/mL in acetonitrile: water

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

solubility

water: soluble

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

single component solution

storage temp.

−20°C

SMILES string

CCCC[C@@H](C)[C@@H](OC(=O)CC(CC(O)=O)C(O)=O)[C@H](C[C@@H](C)C[C@H](O)CCCCCC[C@H](O)[C@@H](C)N)OC(=O)CC(CC(O)=O)C(O)=O

InChI

1S/C34H59NO14/c1-5-6-11-21(3)32(49-31(43)19-24(34(46)47)17-29(40)41)27(48-30(42)18-23(33(44)45)16-28(38)39)15-20(2)14-25(36)12-9-7-8-10-13-26(37)22(4)35/h20-27,32,36-37H,5-19,35H2,1-4H3,(H,38,39)(H,40,41)(H,44,45)(H,46,47)/t20-,21+,22+,23?,24?,25+,26-,27-,32+/m0/s1

InChI key

CPCRJSQNWHCGOP-ZBUYIUKQSA-N

General description

Certan Vial
Fumonisins are mycotoxins, produced by molds of genus Fusarium. Six types of fumonisins and their structures have been discovered: fumonisin (B1, B2, B3, B4, A1 and A2). Fumonisins are responsible for toxic properties of corn contaminated with Fusarium moniliforme. Simultaneous determination of fumonisin B1, fumonisin B2 and fumonisin B3 in traditional chinese medicines by ultra-high-performance liquid chromatography-tandem mass spectrometry is reported.

Application

Fumonisin B3 may be used as a reference standard for the determination of the analyte in food supplements using liquid chromatography coupled with tandem mass spectrometry (LC-MS/MS).
Fumonisin B3 solution was used as analytical standard in determination of fumonisins in stored wheat samples by HPLC analysis. It may be used as analytical standard in determination of fumonisins B1, B2, and B3 and hydrolyzed fumonisin B1 in corn containing foods.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analysis Note

purity : ≥95.0% (HPLC)

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

42.8 °F - not determined

Flash Point(C)

6 °C - not determined

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chuck B Bird et al.
Journal of AOAC International, 85(2), 404-410 (2002-05-07)
Fumonisins-mycotoxins produced by some Fusarium species-have been shown to be the causative agent of diseases in horses and other domesticated animals as well as possible carcinogens in humans. A collaborative study was conducted to evaluate the effectiveness of a competitive
Chenglan Liu et al.
Mycopathologia, 164(3), 127-134 (2007-07-17)
The present work deals with the capability for producing fumonisin by Fusarium proliferatum strains isolated from asparagus in China. Fifty of F. proliferatum strains were randomly selected and incubated on cultures of maize grain and asparagus spear, respectively. Fumonisin levels
L E Broggi et al.
Food additives and contaminants, 19(5), 465-469 (2002-05-25)
Corn samples and different dry-milled fractions collected from an industrial mill in Argentina were analysed. Average contaminations were FB(1) 1540 microg kg(-1), FB(2) 716 microg kg(-1) and FB(3) 152 microg kg(-1) in whole corn; FB(1) 135 microg kg(-1), FB(2) 39.1
M W Trucksess et al.
Food additives and contaminants, 19(7), 671-675 (2002-07-13)
The occurrence of aflatoxins and fumonisins in Incaparina was investigated. Incaparina is a mixture of corn and cottonseed flour with added vitamins, minerals and a preservative. It has been marketed as a high-protein food supplement, particularly for children on protein-deficient
Detection of fumonisins B1, B2, and B3 and hydrolyzed fumonisin B1 in corn-containing foods.
Hopmans EC and Murphy PA.
Journal of Agricultural and Food Chemistry, 41(10), 1655-1658 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service