Skip to Content
Merck
All Photos(1)

Key Documents

T4138

Sigma-Aldrich

HT-2 Toxin

≥98% (HPLC)

Synonym(s):

15-Acetoxy-3α,4β-dihydroxy-8α-(3-methylbutyryloxy)-12,13-epoxytrichothec-9-ene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H32O8
CAS Number:
Molecular Weight:
424.48
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥98% (HPLC)

availability

not available in USA

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

[H][C@]12O[C@]3([H])[C@H](O)[C@@H](O)C(C)([C@@]1(COC(C)=O)C[C@H](OC(=O)CC(C)C)C(C)=C2)C34CO4

InChI

1S/C22H32O8/c1-11(2)6-16(24)29-14-8-21(9-27-13(4)23)15(7-12(14)3)30-19-17(25)18(26)20(21,5)22(19)10-28-22/h7,11,14-15,17-19,25-26H,6,8-10H2,1-5H3/t14-,15+,17+,18+,19+,20?,21+,22?/m0/s1

InChI key

PNKLMTPXERFKEN-CKRUGLOOSA-N

Looking for similar products? Visit Product Comparison Guide

Application

HT-2 toxin is a fusarium mycotoxin (secondary fungal metabolite toxic to human and animals) which may be used as a reference material in systems that detect mycotoxins. HT-2 toxin may be used in in vitro studies to understand its mechanisms of cytotoxicity.

related product

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Benedikt Warth et al.
Rapid communications in mass spectrometry : RCM, 26(13), 1533-1540 (2012-05-29)
Mycotoxins regularly occur in food worldwide and pose serious health risks to consumers. Since individuals can be exposed to a variety of these toxic secondary metabolites of fungi at the same time, there is a demand for proper analytical methods
Francisco M Valle-Algarra et al.
Talanta, 84(4), 1112-1117 (2011-05-03)
There is a need to develop sensitive and accurate analytical methods for determining deoxynivalenol (DON), HT-2 toxin and T-2 toxin in paprika to properly assess the relevant risk of human exposure. An optimized analytical method for determination of HT-2 toxin
S De Baere et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 879(24), 2403-2415 (2011-07-20)
A sensitive and specific method for the quantitative determination of deoxynivalenol (DON), deepoxy-deoxynivalenol (DOM-1), T-2 toxin (T-2) and HT-2 toxin (HT-2) in animal body fluids (plasma and bile) using liquid chromatography combined with electrospray ionization tandem mass spectrometry (LC-ESI-MS/MS) is
Maria Weidner et al.
Journal of agricultural and food chemistry, 60(22), 5676-5684 (2012-05-04)
The mycotoxin T-2 toxin, produced by various Fusarium species, is a widespread contaminant of grain and grain products. Knowledge about its toxicity and metabolism in the human body is crucial for any risk assessment as T-2 toxin can be detected
Linda Monaci et al.
Journal of chromatography. A, 1218(48), 8646-8654 (2011-10-29)
A sensitive and accurate method employing a single stage high resolution mass spectrometer equipped with a high-energy collision-dissociation cell (HCD) for the simultaneous determination of deoxynivalenol (DON), T-2 toxin (T-2) and HT-2 toxin (HT-2) in a processed bread model food

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service